Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
This antibody reacts with human proteinkinase A (an intracellular antigen) catalytic (PKAc) alpha subunit, and weakly with PKAc gamma subunit (both around 40 kDa). The recognized epitope of PKAc alpha is identical between man, sheep, pig, ox and dog.
Protein kinase A (PKA, cAMP-dependent protein kinase) is a key element of a ubiquitous signaling pathway important in the cell cycle, cellular communication, memory formation and behavior. PKA is composed of two catalytic (PKAc; Protein Kinase A catalytic subunit) and two regulatory subunits (PKAr). Upon binding cAMP, the complex dissociates to PKAr dimer and two activated PKAc ser/thr protein kinase catalytic monomers. The released PKAc can translocate into the nucleus and exert a regulatory role in the activation of multiple nuclear hormone receptors. However, PKAc-mediated activation of tonicity-dependent gene expression is cAMP independent. Humans express three types of PKAc subunit - PKAc alpha is present in most human tissues, PKAc beta and gamma are tissue-specific, the later is found in testes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C(s); C-alpha-2; C-alpha-S; cAMP-dependent protein kinase catalytic subunit alpha; cAMP-dependent protein kinase catalytic subunit alpha, isoform 1; MGC102831; MGC48865; PKA; PKA C-alpha; pkac; protein kinase A catalytic subunit; protein kinase, cAMP-dependent, alpha catalytic subunit; protein kinase, cAMP-dependent, catalytic, alpha
Gene Aliases: PKACA; PPNAD4; PRKACA
UniProt ID: (Human) P17612
Entrez Gene ID: (Human) 5566
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support