Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA1-40059 detects JAM-C from mouse and human samples.
MA1-40059 has been successfully used in flow cytometry, immunohistochemistry (frozen), ELISA and immunoprecipitation applications.
The MA1-40059 immunogen is mouse JAM-C.
JAM3 is involved in tight junctions and represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. The protein encoded by this immunoglobulin superfamily gene member is localized in the tight junctions between high endothelial cells. Unlike other proteins in this family, the this protein is unable to adhere to leukocyte cell lines and only forms weak homotypic interactions. The encoded protein is a member of the junctional adhesion molecule protein family and acts as a receptor for another member of this family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: JAM-2; JAM-3; JAM-C; junction cell adhesion molecule 3; junction cell adhesion molecule C; Junctional adhesion molecule 3; Junctional adhesion molecule C
Gene Aliases: 1110002N23Rik; JAM-2; JAM-3; JAM-C; JAM3; JAMC; Jcam3; UNQ859/PRO1868
UniProt ID: (Human) Q9BX67, (Mouse) Q9D8B7
Entrez Gene ID: (Human) 83700, (Mouse) 83964
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support