Product References
The SMYD1 and skNAC transcription factors contribute to neurodegenerative diseases.
Brain, behavior, & immunity - health
Mayfield RD,Zhu L,Smith TA,Tiwari GR,Tucker HO
MA3-042 was used in Immunoprecipitation to report that comparative analysis of genes deregulated by SMYD1 or skNAC knockdown in differentiating C2C12 myoblasts identified transcripts characteristic of neurodegenerative diseases, including Alzheimer's, Parkinson's and Huntington's Diseases (AD, PD, and HD).
Tue Dec 01 00:00:00 EST 2020
Adventitial Cell Atlas of wt (Wild Type) and ApoE (Apolipoprotein E)-Deficient Mice Defined by Single-Cell RNA Sequencing.
Arteriosclerosis, thrombosis, and vascular biology
Gu W,Ni Z,Tan YQ,Deng J,Zhang SJ,Lv ZC,Wang XJ,Chen T,Zhang Z,Hu Y,Jing ZC,Xu Q
MA3-042 was used in Immunohistochemistry-immunofluorescence to study the role of the cellular atlas of the heterogeneous adventitial cells, and the intracellular communication, in atherosclerosis.
Sat Jun 01 00:00:00 EDT 2019
Cross-talk between microglia and neurons regulates HIV latency.
PLoS pathogens
Alvarez-Carbonell D,Ye F,Ramanath N,Garcia-Mesa Y,Knapp PE,Hauser KF,Karn J
Published figure using Acetylcholinesterase monoclonal antibody (Product # MA3-042) in Immunofluorescence
Sun Dec 01 00:00:00 EST 2019
Cross-talk between microglia and neurons regulates HIV latency.
PLoS pathogens
Alvarez-Carbonell D,Ye F,Ramanath N,Garcia-Mesa Y,Knapp PE,Hauser KF,Karn J
Published figure using Acetylcholinesterase monoclonal antibody (Product # MA3-042) in Immunofluorescence
Sun Dec 01 00:00:00 EST 2019
Identification of Carboxylesterase, Butyrylcholinesterase, Acetylcholinesterase, Paraoxonase, and Albumin Pseudoesterase in Guinea Pig Plasma through Nondenaturing Gel Electrophoresis.
Comparative medicine
Napon G,Dafferner AJ,Saxena A,Lockridge O
MA3-042 was used in Immunohistochemistry to identify the major esterases in guinea pig plasma.
Mon Oct 01 00:00:00 EDT 2018
Neuromuscular synapse degeneration without muscle function loss in the diaphragm of a murine model for Huntington's Disease.
Neurochemistry international
Valadão PAC,Gomes MPSM,Aragão BC,Rodrigues HA,Andrade JN,Garcias R,Joviano-Santos JV,Luiz MA,Camargo WL,Naves LA,Kushmerick C,Cavalcante WLG,Gallacci M,de Jesus ICG,Guatimosim S,Guatimosim C
MA3-042 was used in Western Blot to contribute to expanding the current understanding of the effects of mutated huntingtin in the neuromuscular synapse and the diaphragm muscle function.
Fri Jun 01 00:00:00 EDT 2018
Immunopurification of Acetylcholinesterase from Red Blood Cells for Detection of Nerve Agent Exposure.
Chemical research in toxicology
Dafferner AJ,Schopfer LM,Xiao G,Cashman JR,Yerramalla U,Johnson RC,Blake TA,Lockridge O
MA3-042 was used in Western Blotting to develop a method of immunopurifying human red blood cell acetylcholinesterase in quantities adequate for detecting exposure to nerve agents and organophosphorus pesticides, by mass spectrometry.
Mon Oct 16 00:00:00 EDT 2017
Functional variability in butyrylcholinesterase activity regulates intrathecal cytokine and astroglial biomarker profiles in patients with Alzheimer's disease.
Neurobiology of aging
Darreh-Shori T,Vijayaraghavan S,Aeinehband S,Piehl F,Lindblom RP,Nilsson B,Ekdahl KN,Långström B,Almkvist O,Nordberg A
MA3-042 was used in ELISA to study the effect of Alzheimer's disease patient butyrylcholinesterase genotype on the profile of inflammatory cytokines and markers of astroglial function in CSF
Fri Nov 01 00:00:00 EDT 2013
Lesions of cortical GABAergic interneurons and acetylcholine neurons in xeroderma pigmentosum group A.
Brain & development
Hayashi M,Ohto T,Shioda K,Fukatsu R
MA3-042 was used in immunohistochemistry to study the status of cortical GABAergic interneurons and cholinergic neurons in xeroderma pigmentosum group A
Sun Apr 01 00:00:00 EDT 2012
Decrease in acetylcholinergic neurons in the pedunculopontine tegmental nucleus in a patient with Prader-Willi syndrome.
Neuropathology : official journal of the Japanese Society of Neuropathology
Hayashi M,Miyata R,Tanuma N
MA3-042 was used in immunohistochemistry to study the selective deficiency of acetylcholine neurons in the pedunculopontine tegmental nucleus with Prader-Willi syndrome
Wed Jun 01 00:00:00 EDT 2011
Altered levels of acetylcholinesterase in Alzheimer plasma.
PloS one
García-Ayllón MS,Riba-Llena I,Serra-Basante C,Alom J,Boopathy R,Sáez-Valero J
MA3-042 was used in immunoprecipitation to investigate the changes of acetylcholinesterase level in Alzheimer plasma
Thu Jan 14 00:00:00 EST 2010
Altered levels of acetylcholinesterase in Alzheimer plasma.
PloS one
García-Ayllón MS,Riba-Llena I,Serra-Basante C,Alom J,Boopathy R,Sáez-Valero J
MA3-042 was used in immunoprecipitation to investigate the changes of acetylcholinesterase level in Alzheimer plasma
Thu Jan 14 00:00:00 EST 2010
Different cholinesterase inhibitor effects on CSF cholinesterases in Alzheimer patients.
Current Alzheimer research
Nordberg A,Darreh-Shori T,Peskind E,Soininen H,Mousavi M,Eagle G,Lane R
MA3-042 was used in western blot to investigate the effect of cholinesterase inhibitors on the actvities and protein levels of cholinesterases
Sun Feb 01 00:00:00 EST 2009
Presenilin 1 interacts with acetylcholinesterase and alters its enzymatic activity and glycosylation.
Molecular and cellular biology
Silveyra MX,Evin G,Montenegro MF,Vidal CJ,Martínez S,Culvenor JG,Sáez-Valero J
Published figure using Acetylcholinesterase monoclonal antibody (Product # MA3-042) in Immunocytochemistry
Thu May 01 00:00:00 EDT 2008
Presenilin 1 interacts with acetylcholinesterase and alters its enzymatic activity and glycosylation.
Molecular and cellular biology
Silveyra MX,Evin G,Montenegro MF,Vidal CJ,Martínez S,Culvenor JG,Sáez-Valero J
Published figure using Acetylcholinesterase monoclonal antibody (Product # MA3-042) in Immunocytochemistry
Thu May 01 00:00:00 EDT 2008
Presenilin 1 interacts with acetylcholinesterase and alters its enzymatic activity and glycosylation.
Molecular and cellular biology
Silveyra MX,Evin G,Montenegro MF,Vidal CJ,Martínez S,Culvenor JG,Sáez-Valero J
Published figure using Acetylcholinesterase monoclonal antibody (Product # MA3-042) in Immunocytochemistry
Thu May 01 00:00:00 EDT 2008
Fetal fornix transection and gestation length in sheep.
Experimental neurology
McDonald TJ,Li C,Vincent SE,Nijland MJ
MA3-042 was used in immunohistochemistry to study the developmental state of fetal hippocampus and its role in gestation
Tue Aug 01 00:00:00 EDT 2006
Immunocytochemical demonstration of axonal and perikaryal acetylcholinesterase in human cerebral cortex.
Brain research
Mesulam MM,Geula C,Cosgrove R,Mash D,Brimijoin S
MA3-042 was used in immunohistochemistry to investigate the expression of acetylcholinesterase in axons and perikarya in the human cerebral cortex
Fri Jan 25 00:00:00 EST 1991