Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
MA3-942 detects the 80 kDa subunit of m-calpain in human platelets and erythrocytes, bovine platelets, heart and skeletal muscle, rat myoblasts, kidney, liver and spleen, pig cultured cells, and rabbit samples. This antibody does not cross-react with mu-calpain, n-calpain, calmodulin or calpastatin.
MA3-942 has been successfully used in Western blot, IF, immunohistochemistry, and ELISA procedures. By Western blot, this antibody detects an ~80 kDa protein representing m-calpain in bovine platelets. Immunocytochemical staining of m-calpain in LLC-PK1 cells with MA3-942 results in diffuse cytoplasmic staining. This product has not been shown to be effective in immunoprecipitation experiments.
The MA3-942 antigen is purified bovine skeletal muscle 80 kDa subunit of m-calpain. This antibody recognizes an epitope between amino acids 502-699 (domain III/IV) of human calpain.
Mu-Calpain, also known as Calpain-I, and m-calpain, also known as Calpain-II, are intracellular, calcium-dependent cysteine proteases. Mu-calpain has a micromolar sensitivity (thus the mu-) as compared to the millimolar calcium sensitivity of m-calpain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcium-activated neutral proteinase 2; calcium-dependent protease; calpain 2, (m/II) large subunit; calpain 2, large [catalytic] subunit; calpain 2, large subunit; calpain II; Calpain large polypeptide L2; Calpain M-type; calpain, large polypeptide L2; Calpain-2 catalytic subunit; Calpain-2 large subunit; CANP 2; M-calpain; Millimolar-calpain
Gene Aliases: CANP 2; CANP2; CANPL2; CANPml; CAPN2; mCANP
UniProt ID: (Bovine) Q27971, (Human) P17655, (Rat) Q07009
Entrez Gene ID: (Rabbit) 100009092, (Pig) 397393, (Bovine) 281662, (Human) 824, (Rat) 29154
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support