Product References
Characterization of estrogen response element binding proteins as biomarkers of breast cancer behavior.
Clinical biochemistry
Kruer TL,Cummins TD,Powell DW,Wittliff JL
MA5-12325 was used in EMSA to study the prognostic value of estrogen resonse element binding proteins in breast cancer
Fri Nov 01 00:00:00 EDT 2013
Location analysis for the estrogen receptor-alpha reveals binding to diverse ERE sequences and widespread binding within repetitive DNA elements.
Nucleic acids research
Mason CE,Shu FJ,Wang C,Session RM,Kallen RG,Sidell N,Yu T,Liu MH,Cheung E,Kallen CB
MA5-12325 was used in ChIP assay to study the nucleotide sequence binding specificity of estrogen receptor-alpha
Thu Apr 01 00:00:00 EDT 2010
Expression and subcellular distribution of native estrogen receptor beta in murine C2C12 cells and skeletal muscle tissue.
Steroids
Milanesi L,Vasconsuelo A,de Boland AR,Boland R
MA5-12325 was used in western blot to study the expression and subcellular distribution of estrogen receptor beta in murine C2C12 cells and skeletal muscle tissue
Mon Jun 01 00:00:00 EDT 2009
Leukemogenic MLL fusion proteins bind across a broad region of the Hox a9 locus, promoting transcription and multiple histone modifications.
Cancer research
Milne TA,Martin ME,Brock HW,Slany RK,Hess JL
MA5-12325 was used in ChIP assay to study the mechanisms by which leukemogenic MLL proteins induce multiple modifications of histones
Thu Dec 15 00:00:00 EST 2005
Identification of estrogen receptor beta expression in Chinese hamster ovary (CHO) cells and comparison of estrogen-responsive gene transcription in cells adapted to serum-free media.
The Journal of steroid biochemistry and molecular biology
Thomas PB,Risinger KE,Klinge CM
MA5-12325 was used in western blot to study estrogen receptor beta expression in Chinese hamster ovary cells
Tue Jul 01 00:00:00 EDT 2003
Nuclear receptor target gene discovery using high-throughput chromatin immunoprecipitation.
Methods in enzymology
Laganière J,Deblois G,Giguère V
MA5-12325 was used in ChIP assay to identify nuclear receptor target genes by means of chromatin immunoprecipitation
Wed Dec 24 00:00:00 EST 2003
The effects of estrogen, its antagonist ICI 182, 780, and interferon-tau on the expression of estrogen receptors and integrin alphaV beta 3 on cycle day 16 in bovine endometrium.
Reproductive biology and endocrinology : RB&E
Kimmins S,Russell GL,Lim HC,Hall BK,MacLaren LA
MA5-12325 was used in immunohistochemistry to study the effects of estrogen and IFN-tau on the expression of estrogen receptors and integrin alphaV beta 3 in bovine endometrium
Thu Apr 24 00:00:00 EDT 2003
Nuclear receptor function requires a TFTC-type histone acetyl transferase complex.
Molecular cell
Yanagisawa J,Kitagawa H,Yanagida M,Wada O,Ogawa S,Nakagomi M,Oishi H,Yamamoto Y,Nagasawa H,McMahon SB,Cole MD,Tora L,Takahashi N,Kato S
MA5-12325 was used in immunoprecipitation and western blot to study the role of a TFTC-type histone acetyl transferase complex in nuclear receptor function
Fri Mar 01 00:00:00 EST 2002
Occurrence of estrogen receptor alpha in bovine placentomes throughout mid and late gestation and at parturition.
Biology of reproduction
Schuler G,Wirth C,Teichmann U,Failing K,Leiser R,Thole H,Hoffmann B
MA5-12325 was used in immunohistochemistry to study the immunohistochemical expression of estrogen receptor-alpha in bovine placentomes
Mon Apr 01 00:00:00 EST 2002
Affinity chromatography of human estrogen receptor-alpha expressed in Saccharomyces cerevisiae. Combination of heparin- and 17beta-estradiol-affinity chromatography.
Journal of chromatography. A
Feng W,Graumann K,Hahn R,Jungbauer A
MA5-12325 was used in western blot to develop a method for the affinity purification of human estrogen receptor-alpha expressed in Saccharomyces cerevisiae
Fri Aug 06 00:00:00 EDT 1999
Classification of breast cancer cells on the basis of a functional assay for estrogen receptor.
Molecular medicine (Cambridge, Mass.)
Biswas DK,Averboukh L,Sheng S,Martin K,Ewaniuk DS,Jawde TF,Wang F,Pardee AB
MA5-12325 was used in EMSA to study the classification of estrogen receptor-positive breast tumor cells on the basis of a functional estrogen receptor assay
Wed Jul 01 00:00:00 EDT 1998
Identification of a variant estrogen receptor lacking exon 4 and its coexpression with wild-type estrogen receptor in ovarian carcinomas.
Clinical cancer research : an official journal of the American Association for Cancer Research
Park W,Choi JJ,Hwang ES,Lee JH
MA5-12325 was used in western blot to identify and study the expression of a variant estrogen receptor lacking exon 4 in ovarian carcinoma cells
Sun Dec 01 00:00:00 EST 1996