Product References
Optical nanomanipulations of malignant cells: controlled cell damage and fusion.
Small (Weinheim an der Bergstrasse, Germany)
Minai L,Yeheskely-Hayon D,Golan L,Bisker G,Dann EJ,Yelin D
MA5-13045 was used in blocking or activating experiment to study optical nanomanipulations of malignant cells
Mon Jun 11 00:00:00 EDT 2012
Tumor penetration and epidermal growth factor receptor saturation by panitumumab correlate with antitumor activity in a preclinical model of human cancer.
Molecular cancer
Freeman DJ,McDorman K,Ogbagabriel S,Kozlosky C,Yang BB,Doshi S,Perez-Ruxio JJ,Fanslow W,Starnes C,Radinsky R
MA5-13045 was used in immunoprecipitation to study tumor penetration and EGFR saturation by panitumumab and the correlation with therapeutic efficacy in a xenograft model of human epidermoid carcinoma
Wed Jul 25 00:00:00 EDT 2012
A chimeric pre-ubiquitinated EGF receptor is constitutively endocytosed in a clathrin-dependent, but kinase-independent manner.
Traffic (Copenhagen, Denmark)
Bertelsen V,Sak MM,Breen K,Rødland MS,Johannessen LE,Traub LM,Stang E,Madshus IH
MA5-13045 was used in immunocytochemistry and immunohistochemistry to study the roles of EGFR kinase activity and ubiquitination in EGFR endocytosis
Fri Apr 01 00:00:00 EDT 2011
The constitutive activity of epidermal growth factor receptor vIII leads to activation and differential trafficking of wild-type epidermal growth factor receptor and erbB2.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society
Zeineldin R,Ning Y,Hudson LG
MA5-13045 was used in immunocytochemistry to investigate the effect of the constitutive activity of EGFRvIII on wild-type EGFR and erbB2 in OVCA 433 cells
Tue Jun 01 00:00:00 EDT 2010
Ultrastructural characterization of giant endosomes induced by GTPase-deficient Rab5.
Histochemistry and cell biology
Wegner CS,Malerød L,Pedersen NM,Progida C,Bakke O,Stenmark H,Brech A
MA5-13045 was used in flow cytometry to study the ultrastructure of giant endosomes induced by GTPase-deficient Rab5
Fri Jan 01 00:00:00 EST 2010
The juxtamembrane region of the EGF receptor functions as an activation domain.
Molecular cell
Red Brewer M,Choi SH,Alvarado D,Moravcevic K,Pozzi A,Lemmon MA,Carpenter G
MA5-13045 was used in flow cytometry to study the role of the juxtamembrane region of the EGF receptor as an activation domain
Fri Jun 26 00:00:00 EDT 2009
Expression of epidermal growth factor receptor or ErbB3 facilitates geldanamycin-induced down-regulation of ErbB2.
Molecular cancer research : MCR
Pedersen NM,Breen K,Rødland MS,Haslekås C,Stang E,Madshus IH
MA5-13045 was used in flow cytometry to study the interplay among epidermal growth factor receptor, ErbB2, and ErbB3 during the treatment of breast cancer and its therapeutic significance
Sun Feb 01 00:00:00 EST 2009
Prolonged EGFR signaling by ERBB2-mediated sequestration at the plasma membrane.
Traffic (Copenhagen, Denmark)
Offterdinger M,Bastiaens PI
MA5-13045 was used in flow cytometry to study the regulation of EGF receptor signaling by plasma membrane erbB2
Tue Jan 01 00:00:00 EST 2008
Molecular classification system identifies invasive breast carcinoma patients who are most likely and those who are least likely to achieve a complete pathologic response after neoadjuvant chemotherapy.
Cancer
Goldstein NS,Decker D,Severson D,Schell S,Vicini F,Margolis J,Dekhne NS
MA5-13045 was used in immunohistochemistry to investigate the sub-classification invasive breast carcinoma patients and its prognostic significance
Mon Oct 15 00:00:00 EDT 2007
Effects of cisplatin, alpha-interferon, and 13-cis retinoic acid on the expression of Fas (CD95), intercellular adhesion molecule-1 (ICAM-1), and epidermal growth factor receptor (EGFR) in oral cancer cell lines.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology
Sundelin K,Roberg K,Grénman R,Håkansson L
MA5-13045 was used in immunocytochemistry to investigate the effects of chemotherapeutic agents cisplatin, alpha-interferon, and 12-cis retinoic acid on the expression of mutliple proteins in oral cancer cell lines
Thu Mar 01 00:00:00 EST 2007
Activation of the epidermal growth factor (EGF) receptor induces formation of EGF receptor- and Grb2-containing clathrin-coated pits.
Molecular and cellular biology
Johannessen LE,Pedersen NM,Pedersen KW,Madshus IH,Stang E
MA5-13045 was used in immunocytochemistry to study clathrin-coated pits containing EGF, EGFR and Grb2 and the role of EGFR activation in their formation
Sun Jan 01 00:00:00 EST 2006
Both clathrin-positive and -negative coats are involved in endosomal sorting of the EGF receptor.
Experimental cell research
Myromslien FD,Grøvdal LM,Raiborg C,Stenmark H,Madshus IH,Stang E
MA5-13045 was used in immunocytochemistry to study the endosomal sorting of EGFR and the role played by clathrin-positive and clathrin-negative coats
Sun Oct 01 00:00:00 EDT 2006
Novel prognostic value of nuclear epidermal growth factor receptor in breast cancer.
Cancer research
Lo HW,Xia W,Wei Y,Ali-Seyed M,Huang SF,Hung MC
MA5-13045 was used in immunohistochemistry to study the prognostic value of nuclear-localized EGFR in beast cancer
Sat Jan 01 00:00:00 EST 2005
Retention of intrinsic stem cell hierarchies in carcinoma-derived cell lines.
Cancer research
Locke M,Heywood M,Fawell S,Mackenzie IC
MA5-13045 was used in immunocytochemistry to investigate the patterns of colony formation in cell lines derived from carcinomas
Sat Oct 01 00:00:00 EDT 2005
Herceptin-induced inhibition of ErbB2 signaling involves reduced phosphorylation of Akt but not endocytic down-regulation of ErbB2.
International journal of cancer
Longva KE,Pedersen NM,Haslekås C,Stang E,Madshus IH
MA5-13045 was used in immunocytochemistry to study the role of Akt phosphorylation in the inhibition of ErbB2 signaling by Herceptin
Thu Sep 01 00:00:00 EDT 2005
The inhibitory effect of ErbB2 on epidermal growth factor-induced formation of clathrin-coated pits correlates with retention of epidermal growth factor receptor-ErbB2 oligomeric complexes at the plasma membrane.
Molecular biology of the cell
Haslekås C,Breen K,Pedersen KW,Johannessen LE,Stang E,Madshus IH
MA5-13045 was used in immunocytochemistry to study the inhibitory effect of ErbB2 on EGF-induced formation of clathrin-coated pits
Thu Dec 01 00:00:00 EST 2005
Abnormal beta-catenin expression in oral cancer with no gene mutation: correlation with expression of cyclin D1 and epidermal growth factor receptor, Ki-67 labeling index, and clinicopathological features.
Human pathology
Odajima T,Sasaki Y,Tanaka N,Kato-Mori Y,Asanuma H,Ikeda T,Satoh M,Hiratsuka H,Tokino T,Sawada N
MA5-13045 was used in immunohistochemistry to study beta-catenin expression in oral cancer
Tue Mar 01 00:00:00 EST 2005
Expression of the HER1-4 family of receptor tyrosine kinases in breast cancer.
The Journal of pathology
Witton CJ,Reeves JR,Going JJ,Cooke TG,Bartlett JM
MA5-13045 was used in immunohistochemistry to study the expression of HER1-4 in 220 cases of breast carcinoma and the correlation with survival
Tue Jul 01 00:00:00 EDT 2003
Evidence of high incidence of EGFRvIII expression and coexpression with EGFR in human invasive breast cancer by laser capture microdissection and immunohistochemical analysis.
International journal of cancer
Ge H,Gong X,Tang CK
MA5-13045 was used in immunohistochemistry to investigate the incidence of EGFRvIII in human invasive breast cancer
Wed Mar 20 00:00:00 EST 2002
Ligand-independent dimer formation of epidermal growth factor receptor (EGFR) is a step separable from ligand-induced EGFR signaling.
Molecular biology of the cell
Yu X,Sharma KD,Takahashi T,Iwamoto R,Mekada E
MA5-13045 was used in blocking or activating experiment to study ligand-independent dimer formation of epidermal growth factor receptor
Mon Jul 01 00:00:00 EDT 2002