Product References
TNF-α induces vascular endothelial cells apoptosis through overexpressing pregnancy induced noncoding RNA in Kawasaki disease model.
The international journal of biochemistry & cell biology
Jiang C,Fang X,Jiang Y,Shen F,Hu Z,Li X,Huang X
MA5-14000 was used in western blot to analyze a Kawasaki disease model and how TNF alpha induces vascular endothelial cells apoptosis through overexpressing pregnancy induced noncoding RNA
Tue Mar 01 00:00:00 EST 2016
Phosphorylated nerve growth factor-induced clone B (NGFI-B) translocates from the nucleus to mitochondria of stressed rat cardiomyocytes and induces apoptosis.
Stress (Amsterdam, Netherlands)
Xinxing W,Hong F,Rui Z,Yun Z,Jingbo G,Lingjia Q
MA5-14000 was used in western blot to study the effects of stress on the nuclear to mitochondrial translocation of NGFI-B and its role in the induction of cardiomyocyte apoptosis
Sat Sep 01 00:00:00 EDT 2012
Protective effects of trimetazidine on bone marrow mesenchymal stem cells viability in an ex vivo model of hypoxia and in vivo model of locally myocardial ischemia.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban
Xu H,Zhu G,Tian Y
MA5-14000 was used in western blot to study the effect of trimetazidine on the survival of bone marrow mesenchymal stem cells in a model of hypoxia and following transplantation into rat heart
Wed Feb 01 00:00:00 EST 2012
Combined treatment with the Cox-2 inhibitor niflumic acid and PPARγ ligand ciglitazone induces ER stress/caspase-8-mediated apoptosis in human lung cancer cells.
Cancer letters
Kim BM,Maeng K,Lee KH,Hong SH
MA5-14000 was used in western blot to evaluate a combination treatment of niflumic acid and ciglitazone in human lung cancer cells
Fri Jan 28 00:00:00 EST 2011
Sequential caspase-2 and caspase-8 activation is essential for saikosaponin a-induced apoptosis of human colon carcinoma cell lines.
Apoptosis : an international journal on programmed cell death
Kim BM,Hong SH
MA5-14000 was used in immunocytochemistry to investigate the role of caspase activation in the apoptotic effect of saikosaponin in colon carcinoma cell lines
Tue Feb 01 00:00:00 EST 2011
Prenatal stress induces long-term effects in cell turnover in the hippocampus-hypothalamus-pituitary axis in adult male rats.
PloS one
Baquedano E,García-Cáceres C,Diz-Chaves Y,Lagunas N,Calmarza-Font I,Azcoitia I,Garcia-Segura LM,Argente J,Chowen JA,Frago LM
MA5-14000 was used in western blot to study the long-term effects of sub-chronic prenatal stress on cell turnover in the hippocampus-hypothalamus-pituitary axis in adult male rats
Tue Apr 10 00:00:00 EDT 2012
Notch-activated signaling cascade interacts with mitochondrial remodeling proteins to regulate cell survival.
Proceedings of the National Academy of Sciences of the United States of America
Perumalsamy LR,Nagala M,Sarin A
MA5-14000 was used in immunohistochemistry to investigate the interaction between notch signalling and mitochondrial remodeling proteins and its effect on cell survival
Tue Apr 13 00:00:00 EDT 2010
Cell cycle arrest and cytochrome c-mediated apoptotic induction by MCS-5A is associated with up-regulation of p16(INK4a) in HL-60 cells.
Bioorganic & medicinal chemistry letters
Choi BY,Lee CH
MA5-14000 was used in western blot to investigate the effect of cyclin-dependent kinase inhibitor on the cell cycle and apoptosis in HL-60 cells and the underlying mechanism
Thu Jul 01 00:00:00 EDT 2010
Heme oxygenase-1 prevents hyperthyroidism induced hepatic damage via an antioxidant and antiapoptotic pathway.
The Journal of surgical research
Giriş M,Erbil Y,Depboylu B,Mete O,Türkoğlu U,Abbasoğlu SD,Uysal M
MA5-14000 was used in immunohistochemistry to investigate the protective effect of heme oxygenase-1 against hyperthyroidism and its mechanism
Wed Dec 01 00:00:00 EST 2010
N,N-dimethyl phytosphingosine induces caspase-8-dependent cytochrome c release and apoptosis through ROS generation in human leukemia cells.
Toxicology and applied pharmacology
Kim BM,Choi YJ,Han Y,Yun YS,Hong SH
MA5-14000 was used in immunoprecipitation to investigate the mechanism for apoptosis induced by N,N-dimethyl phytosphingosine in human leukemia cells
Sat Aug 15 00:00:00 EDT 2009
Desferrioxamine (DFX) induces apoptosis through the p38-caspase8-Bid-Bax pathway in PHA-stimulated human lymphocytes.
Toxicology and applied pharmacology
Kim BM,Chung HW
MA5-14000 was used in western blot to investigate the effect of desferrioxamine on apoptosis and its mechanism
Tue Apr 01 00:00:00 EDT 2008
Influence of glutathione on the induction of chromosome aberrations, delay in cell cycle kinetics and cell cycle regulator proteins in irradiated mouse bone marrow cells.
International journal of radiation biology
Ray S,Chatterjee A
MA5-14000 was used in western blot to investigate the effect of glutathione on chromosome aberrations, cell cycle kinetics, and cell cycle regulator proteins
Tue May 01 00:00:00 EDT 2007
The N-terminus and alpha-5, alpha-6 helices of the pro-apoptotic protein Bax, modulate functional interactions with the anti-apoptotic protein Bcl-xL.
BMC cell biology
Parikh N,Koshy C,Dhayabaran V,Perumalsamy LR,Sowdhamini R,Sarin A
MA5-14000 was used in immunocytochemistry to investigate the relationship and interaction between Bax and Bcl-xL
Wed May 23 00:00:00 EDT 2007
Iptakalim protects PC12 cell against H2O2-induced oxidative injury via opening mitochondrial ATP-sensitive potassium channel.
Biochemical and biophysical research communications
Chai Y,Niu L,Sun XL,Ding JH,Hu G
MA5-14000 was used in immunocytochemistry to study the protection of PC12 cells against H2O2-induced oxidative injury by iptakalim-mediated opening of mitochondrial ATP-sensitive potassium channels
Fri Nov 17 00:00:00 EST 2006
(+/-)-huprine Y, (-)-huperzine A and tacrine do not show neuroprotective properties in an apoptotic model of neuronal cytoskeletal alteration.
Journal of Alzheimer's disease : JAD
Jordá EG,Verdaguer E,Jiménez A,Canudas AM,Rimbau V,Camps P,Muñoz-Torrero D,Camins A,Pallàs M
MA5-14000 was used in immunocytochemistry to study the effect of acetylcholinesterase inhibitors in colchicine-induced apoptosis in cerebellar granule neurons
Wed Dec 01 00:00:00 EST 2004