Product References
Modulatory Effects of Estradiol and Its Mixtures with Ligands of GPER and PPAR on MAPK and PI3K/Akt Signaling Pathways and Tumorigenic Factors in Mouse Testis Explants and Mouse Tumor Leydig Cells.
Biomedicines
Gorowska-Wojtowicz E,Duliban M,Kotula-Balak M,Bilinska B
MA5-15065 was used in Western Blotting to suggest that a balanced estrogen level and its action together with proper GPER and PPAR signaling play a key role in the maintenance of testis homeostasis.
Sun Jun 12 00:00:00 EDT 2022
LINC02381 aggravates breast cancer through the miR-1271-5p/FN1 axis to activate PI3K/AKT pathway.
Molecular carcinogenesis
Huang S,Huang P,Wu H,Wang S,Liu G
MA5-15065 was used in Western Blotting to conclude that LINC02381 exerts carcinogenic effects in BC by the miR-1271-5p/FN1 axis to activate PI3K/AKT pathway.
Tue Mar 01 00:00:00 EST 2022
The novel anti-colitic effect of β-adrenergic receptors via modulation of PS1/BACE-1/Aβ axis and NOTCH signaling in an ulcerative colitis model.
Frontiers in pharmacology
Nasser S,Abdallah DM,Ahmed KA,Abdel-Mottaleb Y,El-Abhar HS
MA5-15065 was used in Western Blotting to investigate the impact of mirabegron (β3-agonist) and/or carvedilol (β1/β2 antagonist) on iodoacetamide-induced ulcerative colitis with emphasis on the novel pathomechanism of the PS1/BACE-1/Aβ axis in ulcerative colitis, and its relation to the inflammatory cascade, fibrotic processes, and the gut barrier dysfunction.
Fri Nov 18 00:00:00 EST 2022
Dual Targeting of Sorafenib-Resistant HCC-Derived Cancer Stem Cells.
Current oncology (Toronto, Ont.)
Shrestha R,Bridle KR,Cao L,Crawford DHG,Jayachandran A
Published figure using TGF beta-1 monoclonal antibody (Product # MA5-15065) in Western Blot
Fri Jun 11 00:00:00 EDT 2021
Dual Targeting of Sorafenib-Resistant HCC-Derived Cancer Stem Cells.
Current oncology (Toronto, Ont.)
Shrestha R,Bridle KR,Cao L,Crawford DHG,Jayachandran A
Published figure using TGF beta-1 monoclonal antibody (Product # MA5-15065) in Western Blot
Fri Jun 11 00:00:00 EDT 2021
Late Health Effects of Partial Body Irradiation Injury in a Minipig Model Are Associated with Changes in Systemic and Cardiac IGF-1 Signaling.
International journal of molecular sciences
Hritzo B,Aghdam SY,Legesse B,Kaur A,Cao M,Boerma M,Chakraborty N,Dimitrov G,Gautam A,Hammamieh R,Wilkins W,Tsioplaya A,Holmes-Hampton GP,Moroni M
Published figure using TGF beta-1 monoclonal antibody (Product # MA5-15065) in Western Blot
Tue Mar 23 00:00:00 EDT 2021
Combined Inhibition of TGF-β1-Induced EMT and PD-L1 Silencing Re-Sensitizes Hepatocellular Carcinoma to Sorafenib Treatment.
Journal of clinical medicine
Shrestha R,Prithviraj P,Bridle KR,Crawford DHG,Jayachandran A
Published figure using TGF beta-1 monoclonal antibody (Product # MA5-15065) in Immunocytochemistry
Tue Apr 27 00:00:00 EDT 2021
Combined Inhibition of TGF-β1-Induced EMT and PD-L1 Silencing Re-Sensitizes Hepatocellular Carcinoma to Sorafenib Treatment.
Journal of clinical medicine
Shrestha R,Prithviraj P,Bridle KR,Crawford DHG,Jayachandran A
Published figure using TGF beta-1 monoclonal antibody (Product # MA5-15065) in Immunocytochemistry
Tue Apr 27 00:00:00 EDT 2021
Combined Inhibition of TGF-β1-Induced EMT and PD-L1 Silencing Re-Sensitizes Hepatocellular Carcinoma to Sorafenib Treatment.
Journal of clinical medicine
Shrestha R,Prithviraj P,Bridle KR,Crawford DHG,Jayachandran A
Published figure using TGF beta-1 monoclonal antibody (Product # MA5-15065) in Immunocytochemistry
Tue Apr 27 00:00:00 EDT 2021
Role of the Hippo signaling pathway in safflower yellow pigment treatment of paraquat-induced pulmonary fibrosis.
The Journal of international medical research
Li H,Kan B,Song L,Liu Y,Jian X
Published figure using TGF beta-1 monoclonal antibody (Product # MA5-15065) in Western Blot
Tue Sep 01 00:00:00 EDT 2020
Role of the Hippo signaling pathway in safflower yellow pigment treatment of paraquat-induced pulmonary fibrosis.
The Journal of international medical research
Li H,Kan B,Song L,Liu Y,Jian X
Published figure using TGF beta-1 monoclonal antibody (Product # MA5-15065) in Western Blot
Tue Sep 01 00:00:00 EDT 2020
Magnoflorine Ameliorates Inflammation and Fibrosis in Rats With Diabetic Nephropathy by Mediating the Stability of Lysine-Specific Demethylase 3A.
Frontiers in physiology
Chang L,Wang Q,Ju J,Li Y,Cai Q,Hao L,Zhou Y
Published figure using TGF beta-1 monoclonal antibody (Product # MA5-15065) in Western Blot
Mon Jan 11 00:00:00 EST 2021
Magnoflorine Ameliorates Inflammation and Fibrosis in Rats With Diabetic Nephropathy by Mediating the Stability of Lysine-Specific Demethylase 3A.
Frontiers in physiology
Chang L,Wang Q,Ju J,Li Y,Cai Q,Hao L,Zhou Y
Published figure using TGF beta-1 monoclonal antibody (Product # MA5-15065) in Western Blot
Mon Jan 11 00:00:00 EST 2021
Palladin mediates stiffness-induced fibroblast activation in the tumor microenvironment.
Biophysical journal
McLane JS,Ligon LA
MA5-15065 was used in Immunocytochemistry to identify the actin-associated protein palladin as a key node in these signaling pathways that result in fibroblast activation.
Tue Jul 21 00:00:00 EDT 2015
A biomaterial model of tumor stromal microenvironment promotes mesenchymal morphology but not epithelial to mesenchymal transition in epithelial cells.
Acta biomaterialia
McLane JS,Rivet CJ,Gilbert RJ,Ligon LA
MA5-15065 was used in western blot to describe a new in vitro model to test the effect of an aligned fibrous environment on cancer cell morphology and behavior
Sat Nov 01 00:00:00 EDT 2014