Product References
TTF-1/NKX2-1 binds to DDB1 and confers replication stress resistance to lung adenocarcinomas.
Oncogene
Liu Z,Yanagisawa K,Griesing S,Iwai M,Kano K,Hotta N,Kajino T,Suzuki M,Takahashi T
MA516406 was used in western blot to identify interacting proteins of TTF-1 and their role in lung adenocarcinoma cell survival.
Thu Jun 29 00:00:00 EDT 2017
Branchial Cleft-Like Cysts Involving 3 Different Organs: Thyroid Gland, Thymus, and Parotid Gland.
Medicine
Nakazawa T,Kondo T,Oishi N,Tahara I,Kasai K,Inoue T,Mochizuki K,Katoh R
MA5-16406 was used in immunohistochemistry to report a case of multiple cysts arising in both lobes of the thyroid gland, thymus, and right parotid gland
Thu Oct 01 00:00:00 EDT 2015
On the relevance of a testing algorithm for the detection of ROS1-rearranged lung adenocarcinomas.
Lung cancer (Amsterdam, Netherlands)
Mescam-Mancini L,Lantuéjoul S,Moro-Sibilot D,Rouquette I,Souquet PJ,Audigier-Valette C,Sabourin JC,Decroisette C,Sakhri L,Brambilla E,McLeer-Florin A
MA5-16406 was used in immunohistochemistry to evaluate the detection of ROS1 transpositions in lung adenocarcinomas using a testing alogorithm
Sat Feb 01 00:00:00 EST 2014
Influence of thyroid transcription factor-1 on fluorodeoxyglucose uptake and prognosis of non-small cell lung cancer.
Anticancer research
Ooi H,Chen CY,Hsiao YC,Huang WS,Hsieh BT
MA5-16406 was used in immunohistochemistry to study the correlation between fluorodeoxyglucose uptake and thyroid transcription factor-1 expression and the prognostic value in non-small cell lung cancer
Thu May 01 00:00:00 EDT 2014
In search of the ideal immunopanel to distinguish metastatic mammary carcinoma from primary lung carcinoma: a tissue microarray study of 207 cases.
Applied immunohistochemistry & molecular morphology : AIMM
Kawaguchi KR,Lu FI,Kaplan R,Liu YF,Chadwick P,Chen Z,Brogi E,Shin SJ
MA5-16406 was used in immunohistochemistry to establish the optimal panel of immunochemical markers for distinguishing metastatic mammary carcinoma from primary lung carcinoma
Tue Apr 01 00:00:00 EDT 2014
Extracellular matrix defects in aneurysmal Fibulin-4 mice predispose to lung emphysema.
PloS one
Ramnath NW,van de Luijtgaarden KM,van der Pluijm I,van Nimwegen M,van Heijningen PM,Swagemakers SM,van Thiel BS,Ridwan RY,van Vliet N,Vermeij M,Hawinkels LJ,de Munck A,Dzyubachyk O,Meijering E,van der Spek P,Rottier R,Yanagisawa H,Hendriks RW,Kanaar R,Rouwet EV,Kleinjan A,Essers J
MA5-16406 was used in immunohistochemistry - paraffin section to assess lung emphysema and predisposition due to extracellular matrix defects in aneurysmal Fibulin-4 mice
Mon Jun 22 00:00:00 EDT 2015
High-grade lung adenocarcinoma with fetal lung-like morphology: clinicopathologic, immunohistochemical, and molecular analyses of 17 cases.
The American journal of surgical pathology
Morita S,Yoshida A,Goto A,Ota S,Tsuta K,Yokozawa K,Asamura H,Nakajima J,Takai D,Mori M,Oka T,Tamaru J,Itoyama S,Furuta K,Fukayama M,Tsuda H
MA5-16406 was used in immunohistochemistry to report on 17 cases of high-grade lung adenocarcinoma with a fetal lung-like morphology
Sat Jun 01 00:00:00 EDT 2013
Neuroendocrine carcinoma of the stomach: morphologic and immunohistochemical characteristics and prognosis.
The American journal of surgical pathology
Ishida M,Sekine S,Fukagawa T,Ohashi M,Morita S,Taniguchi H,Katai H,Tsuda H,Kushima R
MA5-16406 was used in immunohistochemistry to report on 51 cases of neuroendocrine carcinoma of the stomach
Mon Jul 01 00:00:00 EDT 2013
Adequacy of CT-guided biopsies with histomolecular subtyping of pulmonary adenocarcinomas: influence of ATS/ERS/IASLC guidelines.
Lung cancer (Amsterdam, Netherlands)
Ferretti GR,Busser B,de Fraipont F,Reymond E,McLeer-Florin A,Mescam-Mancini L,Moro-Sibilot D,Brambilla E,Lantuejoul S
MA5-16406 was used in immunohistochemistry to evaluate the quality of pulmonary adenocarcinoma CT-guided trans-thoracic needle biopsies in the context of the 2011 ATS/ERS/IASLC guidelines
Tue Oct 01 00:00:00 EDT 2013
Vimentin expression predicts the occurrence of metastases in non small cell lung carcinomas.
Lung cancer (Amsterdam, Netherlands)
Dauphin M,Barbe C,Lemaire S,Nawrocki-Raby B,Lagonotte E,Delepine G,Birembaut P,Gilles C,Polette M
MA5-16406 was used in immunohistochemistry to study the value of the immunohistochemical expression of vimentin for predicting non-small cell lung carcinoma metastasis
Mon Jul 01 00:00:00 EDT 2013
Extraosseous benign notochordal cell tumor presenting as bilateral pulmonary nodules.
Human pathology
Lee FY,Wen MC,Wang J
MA5-16406 was used in immunohistochemistry to report on a case of extraosseous benign notochordal cell tumor
Mon Jul 01 00:00:00 EDT 2013
Generation of functional thyroid from embryonic stem cells.
Nature
Antonica F,Kasprzyk DF,Opitz R,Iacovino M,Liao XH,Dumitrescu AM,Refetoff S,Peremans K,Manto M,Kyba M,Costagliola S
MA5-16406 was used in immunocytochemistry and immunohistochemistry - paraffin section to report that mouse embryonic stem cells can be used to generate functional thyroid tissue.
Thu Nov 01 00:00:00 EDT 2012
Comparison of monoclonal napsin A, polyclonal napsin A, and TTF-1 for determining lung origin in metastatic adenocarcinomas.
American journal of clinical pathology
Mukhopadhyay S,Katzenstein AL
MA5-16406 was used in immunohistochemistry to perform a comparative study of the value of napsin A and thyroid transcription factor-1 for identifying a pulmonary origin of metastic adenocarcinomas
Thu Nov 01 00:00:00 EDT 2012
Miliary brain metastases from occult lung adenocarcinoma: Radiologic and histopathologic confirmation.
Journal of neurosciences in rural practice
Kahveci R,Gürer B,Kaygusuz G,Sekerci Z
MA5-16406 was used in immunohistochemistry to report on a case of miliary brain metastases from occult lung adenocarcinoma
Sat Sep 01 00:00:00 EDT 2012
A fate map of the murine pancreas buds reveals a multipotent ventral foregut organ progenitor.
PloS one
Angelo JR,Guerrero-Zayas MI,Tremblay KD
MA5-16406 was used in immunohistochemistry to study the presence of a multipotent ventral foregut organ progenitor in mouse pancreatic buds
Thu Mar 21 00:00:00 EDT 2013
Genome-wide analyses of Nkx2-1 binding to transcriptional target genes uncover novel regulatory patterns conserved in lung development and tumors.
PloS one
Tagne JB,Gupta S,Gower AC,Shen SS,Varma S,Lakshminarayanan M,Cao Y,Spira A,Volkert TL,Ramirez MI
MA5-16406 was used in western blot to study transcriptional networks regulated by the Nkx2-1 transcription factor in lung development and tumors
Tue May 08 00:00:00 EDT 2012
Is primary pulmonary meningioma a giant form of a meningothelial-like nodule? A case report and review of the literature.
Case reports in oncology
Masago K,Hosada W,Sasaki E,Murakami Y,Sugano M,Nagasaka T,Yamada M,Yatabe Y
MA5-16406 was used in immunohistochemistry to report on a patient with large pulmonary meningothelial-like nodules
Tue May 01 00:00:00 EDT 2012
The effect of TTF-1 expression on ipsilateral mediastinal nodal (N2) metastases in primary adenocarcinoma of the lung.
Contemporary oncology (Poznan, Poland)
Samancilar O,Kaya SO,Ceylan KC,Usluer O,Yener AG
MA5-16406 was used in immunohistochemistry - paraffin section to examine the effect of thyroid transcription factor-1 expression on ipsilateral mediastinal nodal metastases in primary lung adenocarcinoma
Mon Jun 24 00:00:00 EDT 2013
Specific detection of OCT3/4 isoform A/B/B1 expression in solid (germ cell) tumours and cell lines: confirmation of OCT3/4 specificity for germ cell tumours.
British journal of cancer
Rijlaarsdam MA,van Herk HA,Gillis AJ,Stoop H,Jenster G,Martens J,van Leenders GJ,Dinjens W,Hoogland AM,Timmermans M,Looijenga LH
MA5-16406 was used in immunohistochemistry to study OCT3/4 isoform A/B/B1 expression in solid (germ cell) tumours and cell lines
Tue Sep 06 00:00:00 EDT 2011
Subclassification of non-small cell lung carcinomas lacking morphologic differentiation on biopsy specimens: Utility of an immunohistochemical panel containing TTF-1, napsin A, p63, and CK5/6.
The American journal of surgical pathology
Mukhopadhyay S,Katzenstein AL
MA5-16406 was used in immunohistochemistry to study the utility of an immunohistochemical panel in subtyping non-small cell lung carcinomas lacking morphologic differentiation
Sat Jan 01 00:00:00 EST 2011
Combined small-cell carcinoma of the lung with quadripartite differentiation of epithelial, neuroendocrine, skeletal muscle, and myofibroblastic type.
Virchows Archiv : an international journal of pathology
Pelosi G,Sonzogni A,Galetta D,Perrone F,Braidotti P,Manzotti M,Fabbri A,Spaggiari L,Veronesi G,Viale G
MA5-16406 was used in immunohistochemistry to report on a case of combined small-cell carcinoma of the lung
Fri Apr 01 00:00:00 EDT 2011
Lung-specific inactivation of CCAAT/enhancer binding protein alpha causes a pathological pattern characteristic of COPD.
The European respiratory journal
Didon L,Roos AB,Elmberger GP,Gonzalez FJ,Nord M
MA5-16406 was used in immunohistochemistry to study the role of the transcription factor C/EBPalpha in lung development and its potential role in COPD
Fri Jan 01 00:00:00 EST 2010
TTF1 expression in normal lung neuroendocrine cells and related tumors: immunohistochemical study comparing two different monoclonal antibodies.
Virchows Archiv : an international journal of pathology
La Rosa S,Chiaravalli AM,Placidi C,Papanikolaou N,Cerati M,Capella C
MA5-16406 was used in immunohistochemistry to study the immunohistochemical expression of TTF1 in normal lung neuroendocrine cells and related tumors using two different monoclonal antibodies
Fri Oct 01 00:00:00 EDT 2010
Primary small cell neuroendocrine carcinoma of the kidney: morphological, immunohistochemical, ultrastructural, and cytogenetic study of a case and review of the literature.
Endocrine pathology
La Rosa S,Bernasconi B,Micello D,Finzi G,Capella C
MA5-16406 was used in immunohistochemistry to report on a case of primary small cell neuroendocrine carcinoma of the kidney
Fri Jun 05 00:00:00 EDT 2009
Elevated phospho-S6 expression is associated with metastasis in adenocarcinoma of the lung.
Clinical cancer research : an official journal of the American Association for Cancer Research
McDonald JM,Pelloski CE,Ledoux A,Sun M,Raso G,Komaki R,Wistuba II,Bekele BN,Aldape K
MA5-16406 was used in immunohistochemistry to study the association of elevated phospho-S6 levels with metastasis in adenocarcinoma of the lung
Mon Dec 01 00:00:00 EST 2008
MASH1: a useful marker in differentiating pulmonary small cell carcinoma from Merkel cell carcinoma.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc
Ralston J,Chiriboga L,Nonaka D
MA5-16406 was used in immunohistochemistry to study MASH1 as a marker to differentiate pulmonary small cell carcinoma from Merkel cell carcinoma
Sat Nov 01 00:00:00 EDT 2008
Diagnostic utility of thyroid transcription factors Pax8 and TTF-2 (FoxE1) in thyroid epithelial neoplasms.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc
Nonaka D,Tang Y,Chiriboga L,Rivera M,Ghossein R
MA5-16406 was used in immunohistochemistry to study the diagnostic value of Pax8 and TTF-2 in thyroid epithelial neoplasms
Fri Feb 01 00:00:00 EST 2008
AKR1B10 in usual interstitial pneumonia: expression in squamous metaplasia in association with smoking and lung cancer.
Pathology, research and practice
Li CP,Goto A,Watanabe A,Murata K,Ota S,Niki T,Aburatani H,Fukayama M
MA5-16406 was used in immunohistochemistry to study the role of aldoketo reductase1B10 in the development of lung cancer
Thu Jun 12 00:00:00 EDT 2008
Formation and differentiation of multiple mesenchymal lineages during lung development is regulated by beta-catenin signaling.
PloS one
De Langhe SP,Carraro G,Tefft D,Li C,Xu X,Chai Y,Minoo P,Hajihosseini MK,Drouin J,Kaartinen V,Bellusci S
MA5-16406 was used in immunohistochemistry to study the role of beta-catenin signaling in the formation and differentiation of multiple mesenchymal lineages during lung development
Wed Jan 30 00:00:00 EST 2008
Primary neuroendocrine carcinoma of the breast: a case report.
Tumori
Yaren A,Kelten C,Akbulut M,Teke Z,Duzcan E,Erdem E
MA5-16406 was used in immunohistochemistry to report on a case of primary neuroendocrine carcinoma of the breast
Tue Dec 11 00:00:00 EST 2007
Immunohistopathological re-evaluation of adenocarcinoma of the lung with mixed subtypes using a tissue microarray technique and hierarchical clustering analysis.
Pathology international
Gamal G,Sano T,Sakurai S,Kawashima O,Sugano M,Nakajima T
MA5-16406 was used in immunohistochemistry to study the immunohistopathology of mixed subtypes of adenocarcinoma of the lung
Sat Dec 01 00:00:00 EST 2007
The diagnostic value of TTF-1, CK 5/6, and p63 immunostaining in classification of lung carcinomas.
Applied immunohistochemistry & molecular morphology : AIMM
Kargi A,Gurel D,Tuna B
MA5-16406 was used in immunohistochemistry to study the diagnostic value of TTF-1, CK 5/6, and p63 immunostaining in the classification of lung carcinomas
Sat Dec 01 00:00:00 EST 2007
Prognostic model of stage II non-small cell lung cancer by a discriminant analysis of the immunohistochemical protein expression.
Surgery today
Kohri T,Sugano M,Kawashima O,Saito R,Sakurai S,Sano T,Nakajima T
MA5-16406 was used in immunohistochemistry to develop a prognostic model of non-small cell lung cancer based on protein expression patterns
Thu Nov 01 00:00:00 EDT 2007
The spectrum of pulmonary mucinous cystic neoplasia : a clinicopathologic and immunohistochemical study of ten cases and review of literature.
American journal of clinical pathology
Gao ZH,Urbanski SJ
MA5-16406 was used in immunohistochemistry to study a spectrum of pulmonary mucinous cystic neoplasia cases
Fri Jul 01 00:00:00 EDT 2005
Thyroid transcription factor 1 expression in cystic lesions of the neck: an immunohistochemical investigation of thyroglossal duct cysts, branchial cleft cysts and metastatic papillary thyroid cancer.
Virchows Archiv : an international journal of pathology
Kreft A,Hansen T,Kirkpatrick CJ
MA5-16406 was used in immunohistochemistry to study thyroid transcription factor 1 expression in cystic lesions of the neck
Fri Jul 01 00:00:00 EDT 2005
Pulmonary adenocarcinoma with micropapillary component: an immunohistochemical study. Case report.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica
Kuroda N,Hamauzu T,Toi M,Yamaoka K,Miyazaki E,Hiroi M,Nakata H,Taguchi H,Enzan H
MA5-16406 was used in immunohistochemistry to report on a patient with pulmonary adenocarcinoma with a micropapillary component
Tue Sep 13 00:00:00 EDT 2005
Papillary adenocarcinoma of the lung is a more advanced adenocarcinoma than bronchioloalveolar carcinoma that is composed of two distinct histological subtypes.
Pathology international
Jian Z,Tomizawa Y,Yanagitani N,Iijima H,Sano T,Nakajima T
MA5-16406 was used in immunohistochemistry to study the histology of papillary adenocarcinoma of the lung
Sat Oct 01 00:00:00 EDT 2005
CDX-2 homeobox gene product expression in neuroendocrine tumors: its role as a marker of intestinal neuroendocrine tumors.
The American journal of surgical pathology
Barbareschi M,Roldo C,Zamboni G,Capelli P,Cavazza A,Macri E,Cangi MG,Chilosi M,Doglioni C
MA5-16406 was used in immunohistochemistry to study the expression of the CDX-2 homeobox gene product in neuroendocrine tumors
Wed Sep 01 00:00:00 EDT 2004
TTF-1 and surfactant-B as co-adjuvants in the diagnosis of lung adenocarcinoma and pleural mesothelioma.
Annals of diagnostic pathology
Bakir K,Koçer NE,Deniz H,Güldür ME
MA5-16406 was used in immunohistochemistry to study the diagnostic value of TTF-1 and surfactant-B in lung adenocarcinoma and pleural mesothelioma
Wed Dec 01 00:00:00 EST 2004
Cell type-specific occurrence of caveolin-1alpha and -1beta in the lung caused by expression of distinct mRNAs.
The Journal of biological chemistry
Kogo H,Aiba T,Fujimoto T
MA5-16406 was used in immunohistochemistry to study the role of distinct mRNAs in the cell type-specific occurrence of caveolin-1alpha and -1beta in the lung
Fri Jun 11 00:00:00 EDT 2004
Expression of thyroid transcription factor-1 in 16 human lung cancer cell lines.
Lung cancer (Amsterdam, Netherlands)
Fujita J,Ohtsuki Y,Bandoh S,Ueda Y,Kubo A,Tojo Y,Yamaji Y,Ishida T
MA5-16406 was used in immunocytochemistry to study the expression of thyroid transcription factor-1 in 16 human lung cancer cell lines
Wed Jan 01 00:00:00 EST 2003
Placental transmogrification of the lung is a histologic pattern frequently associated with pulmonary fibrochondromatous hamartoma.
Archives of pathology & laboratory medicine
Xu R,Murray M,Jagirdar J,Delgado Y,Melamed J
MA5-16406 was used in immunohistochemistry to study the association of placental transmogrification of the the lung with pulmonary fibrochondromatous hamartoma
Wed May 01 00:00:00 EDT 2002
The WNT7b promoter is regulated by TTF-1, GATA6, and Foxa2 in lung epithelium.
The Journal of biological chemistry
Weidenfeld J,Shu W,Zhang L,Millar SE,Morrisey EE
MA5-16406 was used in EMSA to study the regulation of WNT7b by TTF-1, GATA6, and Foxa2 in lung epithelium
Fri Jun 07 00:00:00 EDT 2002
A model of pulmonary adenocarcinoma in transgenic mice expressing the simian virus 40 T antigen driven by the rat Calbindin-D9K (CaBP9K) promoter.
The Journal of pathology
Chailley-Heu B,Rambaud C,Barlier-Mur AM,Galateau-Salle F,Perret C,Capron F,Lacaze-Masmonteil T
MA5-16406 was used in immunohistochemistry to develop a transgenic mouse model of pulmonary adenocarcinoma
Thu Nov 01 00:00:00 EST 2001