Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-17188 targets THPO in FACS, indirect ELISA, and WB applications and shows reactivity with Human samples.
The MA5-17188 immunogen is purified recombinant fragment of human THPO expressed in E. Coli.
MA5-17188 detects THPO which has a predicted molecular weight of approximately 37.8kDa.
Thrombopoietin (TPO, THPO, MPL ligand, megakaryocyte colony-stimulating factor) is responsible for the generation of platelets through its action on hematopoietic cells committed to the megakaryocyte lineage. Binding of thrombopoietin to its receptor, MPL (CD110), initiates a signaling cascade via JAK/STAT, MAPK, and PI3K/AKT signaling pathways that leads to the proliferation and differentiation of megakaryocytes and, ultimately, to the production of platelets (thrombopoiesis). Thrombopoietin is constitutively produced by the liver and has a predicted molecular weight of 38 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-mpl ligand; Megakaryocyte colony-stimulating factor; Megakaryocyte growth and development factor; megakaryocyte stimulating factor; MGDF; ML; MPL ligand; Myeloproliferative leukemia virus oncogene ligand; prepro-thrombopoietin; Thrombopoietin
Gene Aliases: MGDF; MKCSF; ML; MPLLG; THCYT1; THPO; TPO
UniProt ID: (Human) P40225
Entrez Gene ID: (Human) 7066
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support