Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-17727 targets CD70/Ki-24 antigen in FACS and IHC (F) applications and shows reactivity with Human samples.
The MA5-17727 immunogen is mouse monoclonal antibodies raised against human CD70.
The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for TNFRSF27/CD27. It is a surface antigen on activated, but not on resting, T and B lymphocytes. It induces proliferation of costimulated T cells, enhances the generation of cytolytic T cells, and contributes to T cell activation. This cytokine is also reported to play a role in regulating B-cell activation, cytotoxic function of natural killer cells, and immunoglobulin synthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD27 ligand; CD27-L; CD70; CD70 antigen; Ki-24 antigen; surface antigen CD70; tumor necrosis factor (ligand) superfamily, member 7; tumor necrosis factor ligand 8A; Tumor necrosis factor ligand superfamily member 7
Gene Aliases: CD27L; CD27LG; CD70; TNFSF7; TNLG8A
UniProt ID: (Human) P32970
Entrez Gene ID: (Human) 970
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support