Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: AVMLVIFYKM RKQHHRQNHH APTRTVEIIN VDDEITGDTP MESHLPMPAI EHEHLNHYNS YKSPFNHTTT VNTINSIHSS VHEPLLIRMN SKDNVQETQI
NGL1 is a specific binding partner for netrin G1 (NTNG1), which is a member of the netrin family of axon guidance molecules (Lin et al., 2003).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Leucine-rich repeat-containing protein 4C; Netrin-G1 ligand; NGL-1
Gene Aliases: KIAA1580; LRRC4C; NGL-1; NGL1; UNQ292/PRO331
UniProt ID: (Human) Q9HCJ2
Entrez Gene ID: (Human) 57689
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support