Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
In ELISAs, this antibody does not cross-react or interfere recombinant human (rh) CXCL/PF4, recombinant mouse (rm) CXCL9/MIG, rmGM-CSF, rmCXCL11/I-TAC, rhCXCL8/IL-8, rmIL-5, or rhCXCL10/IP-10. In Western blots, no cross-reactivity was observed with rhGCP-2, rmGCP-2, rhGRO alpha, rhGRO beta, rhGRO gamma, rhMIP-1 alpha, rmMIP-1 alpha, rmMIP-1 beta, rhMIP-1 gamma, rmMIP-1 gamma, rmMIP-2, rhMIP-3 alpha, rmMIP-3 alpha, rrMIP-3 alpha, rhMIP-3 beta, or rmMIP-3 beta,
Reconstitute at 0.5 mg/mL in sterile PBS.
Platelet factor-4 is a 70-amino acid protein that is released from the alpha-granules of activated platelets and binds with high affinity to heparin. Its major physiologic role appears to be neutralization of heparin-like molecules on the endothelial surface of blood vessels, thereby inhibiting local antithrombin III activity and promoting coagulation. As a strong chemoattractant for neutrophils and fibroblasts, PF4 probably has a role in inflammation and wound repair (Eisman et al., 1990).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-X-C motif chemokine 4; chemokine (C-X-C motif) ligand 4; PF-4; Platelet factor 4
Gene Aliases: Cxcl4; Pf4; Scyb4
UniProt ID: (Mouse) Q9Z126
Entrez Gene ID: (Mouse) 56744
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support