Product References
Development of a skin- and neuro-attenuated live vaccine for varicella.
Nature communications
Wang W,Pan D,Fu W,Ye X,Han J,Yang L,Jia J,Liu J,Zhu R,Zhang Y,Liu C,Ye J,Selariu A,Que Y,Zhao Q,Wu T,Li Y,Zhang J,Cheng T,Zhu H,Xia N
MHCD1401 was used in Flow Cytometry to design a live-attenuated varicella vaccine candidate, v7D.
Fri Feb 11 00:00:00 EST 2022
Development of a skin- and neuro-attenuated live vaccine for varicella.
Nature communications
Wang W,Pan D,Fu W,Ye X,Han J,Yang L,Jia J,Liu J,Zhu R,Zhang Y,Liu C,Ye J,Selariu A,Que Y,Zhao Q,Wu T,Li Y,Zhang J,Cheng T,Zhu H,Xia N
MHCD1401 was used in Flow Cytometry to design a live-attenuated varicella vaccine candidate, v7D.
Fri Feb 11 00:00:00 EST 2022
Mesenchymal stem cells negatively regulate CD4+ T cell activation in patients with primary Sjögren syndrome through the miRNA‑125b and miRNA‑155 TCR pathway.
Molecular medicine reports
Gong B,Zheng L,Lu Z,Huang J,Pu J,Pan S,Zhang M,Liu J,Tang J
MHCD1401 was used in Flow cytometry/Cell sorting to indicate that the expression levels of miRNA‚125b‚5p and miRNA‚155 in TCR‚activated CD4+ T cells from patients with pSS may provide insight regarding autoimmune diseases and offer a novel target for prospective treatment.
Fri Jan 01 00:00:00 EST 2021
Human Tumor-Associated Macrophage and Monocyte Transcriptional Landscapes Reveal Cancer-Specific Reprogramming, Biomarkers, and Therapeutic Targets.
Cancer cell
Cassetta L,Fragkogianni S,Sims AH,Swierczak A,Forrester LM,Zhang H,Soong DYH,Cotechini T,Anur P,Lin EY,Fidanza A,Lopez-Yrigoyen M,Millar MR,Urman A,Ai Z,Spellman PT,Hwang ES,Dixon JM,Wiechmann L,Coussens LM,Smith HO,Pollard JW
MHCD1401 was used in Flow cytometry/Cell sorting to show that monocyte subpopulation distribution and transcriptomes are significantly altered by the presence of endometrial and breast cancer.
Mon Apr 15 00:00:00 EDT 2019
Biomechanical Stability and Osteogenesis in a Tibial Bone Defect Treated by Autologous Ovine Cord Blood Cells-A Pilot Study.
Molecules (Basel, Switzerland)
Herten M,Zilkens C,Thorey F,Tassemeier T,Lensing-Höhn S,Fischer JC,Sager M,Krauspe R,Jäger M
MHCD1401 was used in Flow Cytometry to elucidate the impact of autologous umbilical cord blood cells on bone regeneration and biomechanical stability in an ovine tibial bone defect.
Tue Jan 15 00:00:00 EST 2019
Alterations in the properties of the cell membrane due to glycosphingolipid accumulation in a model of Gaucher disease.
Scientific reports
Batta G,Soltész L,Kovács T,Bozó T,Mészár Z,Kellermayer M,Szöllősi J,Nagy P
MHCD1401-4 was used in Flow cytometry to imply that glycosphingolipid accumulation in the plasma membrane has significant effects on membrane properties, which may be important in the pathogenesis of Gaucher disease.
Tue Jan 09 00:00:00 EST 2018
CD14 and IL18 gene polymorphisms associated with colorectal cancer subsite risks among atomic bomb survivors.
Human genome variation
Hu Y,Yoshida K,Cologne JB,Maki M,Morishita Y,Sasaki K,Hayashi I,Ohishi W,Hida A,Kyoizumi S,Kusunoki Y,Tokunaga K,Nakachi K,Hayashi T
Published figure using CD14 monoclonal antibody (Product # MHCD1401) in Flow Cytometry
Fri Apr 15 00:00:00 EDT 2016
CD14 and IL18 gene polymorphisms associated with colorectal cancer subsite risks among atomic bomb survivors.
Human genome variation
Hu Y,Yoshida K,Cologne JB,Maki M,Morishita Y,Sasaki K,Hayashi I,Ohishi W,Hida A,Kyoizumi S,Kusunoki Y,Tokunaga K,Nakachi K,Hayashi T
Published figure using CD14 monoclonal antibody (Product # MHCD1401) in Flow Cytometry
Fri Apr 15 00:00:00 EDT 2016
Regulation of osteoarthritis-associated key mediators by TNFα and IL-10: effects of IL-10 overexpression in human synovial fibroblasts and a synovial cell line.
Cell and tissue research
Mrosewski I,Jork N,Gorte K,Conrad C,Wiegand E,Kohl B,Ertel W,John T,Oberholzer A,Kaps C,Schulze-Tanzil G
MHCD1401 was used in immunocytochemistry to investigate the effect of TNF and IL-10 on synovial fibroblasts
Tue Jul 01 00:00:00 EDT 2014
Transmission of HIV-1 by primary human uterine epithelial cells and stromal fibroblasts.
The Journal of infectious diseases
Asin SN,Fanger MW,Wildt-Perinic D,Ware PL,Wira CR,Howell AL
MHCD1401 was used in Flow cytometry/Cell sorting to characterise the life-cycle events of human immunodeficiency virus type 1 in primary cultures of human uterine epithelial cells and stromal fibroblasts.
Thu Jul 15 00:00:00 EDT 2004
Susceptibility of immature and mature Langerhans cell-type dendritic cells to infection and immunomodulation by human cytomegalovirus.
Journal of virology
Hertel L,Lacaille VG,Strobl H,Mellins ED,Mocarski ES
MHCD1401 was used in flow cytometry to report that CD34(+) progenitor cell-derived Langerhans cells-type DCs exhibit a differentiation state-dependent susceptibility to CMV infection.
Tue Jul 01 00:00:00 EDT 2003
Flow cytometry in the bone marrow staging of mature B-cell neoplasms.
Cytometry. Part B, Clinical cytometry
Stacchini A,Demurtas A,Godio L,Martini G,Antinoro V,Palestro G
MHCD1401 was used in flow cytometry to ask if flow cytometry can be used for the detection of bone marrow involvement in the different types of B-cell malignant neoplasms.
Tue Jul 01 00:00:00 EDT 2003
Latent cytomegalovirus down-regulates major histocompatibility complex class II expression on myeloid progenitors.
Blood
Slobedman B,Mocarski ES,Arvin AM,Mellins ED,Abendroth A
MHCD1401 was used in Flow Cytometry to study the mechanism involving the modulation of major histocompatibility complex class I and II proteins.
Tue Oct 15 00:00:00 EDT 2002
Dependence of bacterial protein adhesins on toll-like receptors for proinflammatory cytokine induction.
Clinical and diagnostic laboratory immunology
Hajishengallis G,Martin M,Sojar HT,Sharma A,Schifferle RE,DeNardin E,Russell MW,Genco RJ
MHCD1401 was used in flow cytometry to determine of proinflammatory cytokine responses induced by certain bacterial protein adhesins are dependent on TLRs.
Fri Mar 01 00:00:00 EST 2002
Cytokine expression, natural killer cell activation, and phenotypic changes in lymphoid cells from rhesus macaques during acute infection with pathogenic simian immunodeficiency virus.
Journal of virology
Giavedoni LD,Velasquillo MC,Parodi LM,Hubbard GB,Hodara VL
MHCD1401 was used in flow cytometry to examine the innate and adaptive immune responses of rhesus macaques infected with SIVmac251.
Tue Feb 01 00:00:00 EST 2000