Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MM500DB targets Mouse GM-CSF in WB applications and shows reactivity with mouse samples.
The MM500DB immunogen is recombinant mouse GM-CSF.
MM500DB detects Mouse GM-CSF which has a predicted molecular weight of approximately 14 kDa.
The MM500DB mouse GMCSF antibody (biotinylated conjugate of clone 31G6) has successfully been paired as the detection antibody in a sandwich ELISA with capture antibody MM500C (clone 22E9) Typical dilutions for sandwich ELISA include 1 µg/mL for coating and 0.125 - 0.25 µg/mL for detection.
Biotinylated antibody MM500DB (clone 31G6) and antibody MM500C (clone 22E9) have successfully been used in combination with recombinant mouse GMCSF protein SMGMCSF in ELISA applications.
This antibody is produced by injecting Rat IgG secreting hybridoma cells into the peritoneum of mice. The resulting ascites is collected from the mice and the antibody is purified.
The GM-CSF gene encodes for a cytokine that regulates the production, differentiation, and function of macrophages and granulocytes. The active form of the protein exists as a homodimer in the extracellular space. This gene is located in a cluster of related genes at the chromosome region 5q31, which has been associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include interleukins 4, 5, and 13. The gene is involved in promoting tissue inflammation. Elevated levels of cytokines, including the one produced by this gene, have been observed in SARS-CoV-2 infected patients with acute respiratory distress syndrome. Mice that lack this gene or its receptor have been shown to develop pulmonary alveolar proteinosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Colony-stimulating factor; CSF; GM-CSF; granulocyte macrophage colony stimulating factor; granulocyte-macrophage colony stimulating factor 2; Granulocyte-macrophage colony-stimulating factor; M-GM-CSF; MGC131935; MGC138897; MGC151255; MGC151257; put. GM-CSF
Gene Aliases: Csf2; Csfgm; Gm-CSf; GMCSF; MGI-IGM
UniProt ID: (Mouse) P01587
Entrez Gene ID: (Mouse) 12981
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support