Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This antibody is not recommended for ICC/IF applications.
Suggested positive control: mouse kidney lysate or protein.
This gene encodes a member of the WNK family of serine-threonine protein kinases. The kinase is part of the tight junction complex in kidney cells, and regulates the balance between NaCl reabsorption and K(+) secretion. The kinase regulates the activities of several types of ion channels, cotransporters, and exchangers involved in electrolyte flux in epithelial cells. Mutations in this gene result in pseudohypoaldosteronism type IIB.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein kinase lysine-deficient 4; Protein kinase with no lysine 4; protein kinase, lysine deficient 4; protein kinase, lysine-deficient 4; RP23-281C18.8; Serine/threonine-protein kinase WNK4; WNK4 Ser/Thr kinase
Gene Aliases: 2010002J11Rik; Ac2-059; PHA2B; PRKWNK4; WNK4
UniProt ID: (Human) Q96J92, (Mouse) Q80UE6, (Rat) Q7TPK6
Entrez Gene ID: (Human) 65266, (Mouse) 69847, (Rat) 287715
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support