Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PA1-4202 detects CD100 in human samples.
PA1-4202 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~158 kDa protein representing CD100 in transfected CHO cells.
The PA1- 4202 immunogen is a synthetic peptide corresponding to residues K(777) P K S D F C D R E Q S(788) of human CD100.
Semaphorins are neuronal chemorepellants that direct pioneering neurons during nervous system development. CD100 (also known as semaphorin 4d), a 150 kDa homodimer, is a novel semaphorin that induces B cells to aggregate and improves their viability in vitro. CD100 modifies CD40-CD40L B cell signaling by augmenting B cell aggregation and survival and down regulating CD23 expression. CD72 is a lymphocyte receptor for the class IV semaphorin CD100, which is involved in regulating B cell signaling. CD100 is expressed on resting and PHA stimulated T cells. The protein is weakly expressed on NK cells, EBV transformed B cells, monocytes and tumor/peripheral blood T cell lines, and at higher density on activated T cells and B cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A8; BB18; CD100; GR3; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4D; Semaphorin-4D
Gene Aliases: C9orf164; CD100; coll-4; M-sema-G; SEMA4D; SEMAJ
UniProt ID: (Human) Q92854
Entrez Gene ID: (Human) 10507
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support