Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QQQGLQTVDI RAQLLIGAGN SSQSTHMTQR DQQEVIQKFQ DGTLNLLVAT SVAEEGLDIP HCNVVVRYGL LTNEISMVQA RGRARADQSV YAFVATEGSR ELKRELINEA LETLMEQAVA AVQKMDQAEY QAKIRDLQQA ALTKRAAQAA QRENQRQQFP VEHVQLLCIN CMVAVGHGSD LRKVEGTHHV NVNPNFSNYY NVSRDPVVIN KVFKDWKPGG VISCRNCGEV WGLQMIYKSV KLPVLKVRSM LLETPQGRIQ AKKWSRVPFS VPDFDFLQHC AENLSDLSLD
Anti-viral innate immune responses are triggered by pathogen-associated molecular patters (such as the accumulation of intracellular nucleic acids resulting from virus infections) and represent the first line of defense against numerous infectious organisms. The Toll-like receptors (TLR) 3, 7, 8, and 9 are expressed in immune cells and function as transmembrane pattern recognition receptors to detect foreign nucleic acids. Other proteins that play similar roles, such as RIG-1 and MDA5 are members of a CARD-helicase family and are expressed in the cytoplasm. A third protein, LGP2, is similar to RIG-1 and MDA5, except for lacking a homologous CARD domain. It is thought to act as an element of negative-feedback regulation of intracellular antiviral signaling. At least four isoforms of LGP2 are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent helicase LGP2; ATP-dependent RNA helicase DHX58; DEXH (Asp-Glu-X-His) box polypeptide 58; ortholog of mouse D11lgp2; probable ATP-dependent helicase LGP2; probable ATP-dependent RNA helicase DHX58; Protein D11Lgp2; Protein D11Lgp2 homolog; RIG-I-like receptor 3; RIG-I-like receptor LGP2; RLR; RLR-3; RNA helicase LGP2
Gene Aliases: B430001I08Rik; D11LGP2; D11LGP2E; DHX58; LGP2; LPG2; RGD1310093; RLR-3
UniProt ID: (Human) Q96C10, (Mouse) Q99J87
Entrez Gene ID: (Human) 79132, (Mouse) 80861, (Rat) 303538
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support