Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AGDTHCPAEP LAREGTLWEA LRALLPHSKE DLKLDLGEKV ERSVVTLLQR
For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
Predicted homology: Cow: 83%; Guinea Pig: 100%; Horse: 83%; Human: 100%; Mouse: 91%; Rat: 82%
This gene likely encodes a histone lysine demethylase. Studies of a similar protein in mouse indicate a potential role for this protein as a tumor suppressor. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bifunctional peptidase and arginyl-hydroxylase JMJD5; JmjC domain-containing protein 5; Jumonji C domain-containing protein 5; jumonji domain containing 5; jumonji domain-containing protein 5; L-arginine (3R)-hydroxylase KDM8; lysine (K)-specific demethylase 8
Gene Aliases: JMJD5; KDM8
UniProt ID: (Human) Q8N371
Entrez Gene ID: (Human) 79831
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support