Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VDITSKGKMR ARYVNYIKTS EVVRLPYPLQ MKSSGPPSYF IKRESWGWTD
For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
EMC7 (ER Membrane Protein Complex Subunit 7) is a Protein Coding gene. Diseases associated with EMC7 include Cone-Rod Dystrophy, X-Linked, 1. Gene Ontology (GO) annotations related to this gene include carbohydrate binding. [GeneCards]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chromosome 15 hypothetical ATG/GTP binding protein; Endoplasmic reticulum membrane protein complex subunit 7; ER membrane protein complex subunit 7; UPF0480 protein C15orf24
Gene Aliases: C11orf3; C15orf24; EMC7; HT022; ORF1-FL1; UNQ905/PRO1926
UniProt ID: (Human) Q9NPA0
Entrez Gene ID: (Human) 56851
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support