Product References
Downregulation of miR-375 contributes to ERBB2-mediated VEGFA overexpression in esophageal cancer.
Journal of Cancer
Ren S,Tan X,Fu MZ,Ren S,Wu X,Chen T,Latham PS,Lin P,Man YG,Fu SW
Published figure using VEGF polyclonal antibody (Product # PA5-16754) in Western Blot
Fri Nov 05 00:00:00 EDT 2021
Flunixin Meglumine Enhanced Bone Fracture Healing in Rabbits Associated with Activation of Early Collagen Deposition and Enhancement of Vascular Endothelial Growth Factor Expression.
Animals : an open access journal from MDPI
Elgendy M,Elsayad G,Seleim M,Abdo W,Baty RS,Elmahallawy EK,Atiba A
Published figure using VEGF polyclonal antibody (Product # PA5-16754) in Immunohistochemistry
Tue Sep 28 00:00:00 EDT 2021
Triptolide decreases tumor-associated macrophages infiltration and M2 polarization to remodel colon cancer immune microenvironment via inhibiting tumor-derived CXCL12.
Journal of cellular physiology
Jiang X,Cao G,Gao G,Wang W,Zhao J,Gao C
Published figure using VEGF polyclonal antibody (Product # PA5-16754) in Western Blot
Fri Jan 01 00:00:00 EST 2021
Coumestrol mitigates retinal cell inflammation, apoptosis, and oxidative stress in a rat model of diabetic retinopathy via activation of SIRT1.
Aging
Xu Y,Zhang Y,Liang H,Liu X
Published figure using VEGF polyclonal antibody (Product # PA5-16754) in Western Blot
Mon Feb 01 00:00:00 EST 2021
The protective effect of Ganoderma lucidum on testicular torsion/detorsion-induced ischemia-reperfusion (I/R) injury.
Acta cirurgica brasileira
Doğan G,İpek H
Published figure using VEGF polyclonal antibody (Product # PA5-16754) in Immunohistochemistry
Fri May 01 00:00:00 EDT 2020
Hypoxia and proangiogenic proteins in human ameloblastoma.
Scientific reports
de Mendonça RP,Balbinot KM,Martins BV,da Silva Kataoka MS,Mesquita RA,de Jesus Viana Pinheiro J,de Melo Alves Júnior S
PA5-16754 was used in Immunohistochemistry to investigate the expression of the HIF-1α, MMP-2, VEGF, and VEGFR-2 proteins among 24 ameloblastoma cases, 10 calcifying odontogenic cysts, and 9 dental follicles by immunohistochemistry.
Fri Oct 16 00:00:00 EDT 2020
Hypoxia and proangiogenic proteins in human ameloblastoma.
Scientific reports
de Mendonça RP,Balbinot KM,Martins BV,da Silva Kataoka MS,Mesquita RA,de Jesus Viana Pinheiro J,de Melo Alves Júnior S
PA5-16754 was used in Immunohistochemistry to investigate the expression of the HIF-1α, MMP-2, VEGF, and VEGFR-2 proteins among 24 ameloblastoma cases, 10 calcifying odontogenic cysts, and 9 dental follicles by immunohistochemistry.
Fri Oct 16 00:00:00 EDT 2020
Androgen Deprivation Induces Reprogramming of Prostate Cancer Cells to Stem-Like Cells.
Cells
Sánchez BG,Bort A,Vara-Ciruelos D,Díaz-Laviada I
PA5-16754 was used in Western Blotting to provide a new regulatory mechanism of prostate cancer plasticity through AMPK that is worth exploring.
Wed Jun 10 00:00:00 EDT 2020
Androgen Deprivation Induces Reprogramming of Prostate Cancer Cells to Stem-Like Cells.
Cells
Sánchez BG,Bort A,Vara-Ciruelos D,Díaz-Laviada I
PA5-16754 was used in Western Blotting to provide a new regulatory mechanism of prostate cancer plasticity through AMPK that is worth exploring.
Wed Jun 10 00:00:00 EDT 2020
microRNA-21-5p dysregulation in exosomes derived from heart failure patients impairs regenerative potential.
The Journal of clinical investigation
Qiao L,Hu S,Liu S,Zhang H,Ma H,Huang K,Li Z,Su T,Vandergriff A,Tang J,Allen T,Dinh PU,Cores J,Yin Q,Li Y,Cheng K
PA5-16754 was used in Western Blotting to suggest miR-21-5p contributes to exosome-mediated heart repair by enhancing angiogenesis and cardiomyocyte survival through the phosphatase and tensin homolog/Akt pathway.
Mon Apr 29 00:00:00 EDT 2019
microRNA-21-5p dysregulation in exosomes derived from heart failure patients impairs regenerative potential.
The Journal of clinical investigation
Qiao L,Hu S,Liu S,Zhang H,Ma H,Huang K,Li Z,Su T,Vandergriff A,Tang J,Allen T,Dinh PU,Cores J,Yin Q,Li Y,Cheng K
PA5-16754 was used in Western Blotting to suggest miR-21-5p contributes to exosome-mediated heart repair by enhancing angiogenesis and cardiomyocyte survival through the phosphatase and tensin homolog/Akt pathway.
Mon Apr 29 00:00:00 EDT 2019
Fat-Specific Protein 27 Regulation of Vascular Function in Human Obesity.
Journal of the American Heart Association
Karki S,Farb MG,Sharma VM,Jash S,Zizza EJ,Hess DT,Carmine B,Carter CO,Pernar LI,Apovian CM,Puri V,Gokce N
PA5-16754 was used in Western Blotting to support a key role and functional significance of FSP27 as a critical endogenous modulator of human microvascular function that has not been previously described.
Tue Jun 04 00:00:00 EDT 2019
Fat-Specific Protein 27 Regulation of Vascular Function in Human Obesity.
Journal of the American Heart Association
Karki S,Farb MG,Sharma VM,Jash S,Zizza EJ,Hess DT,Carmine B,Carter CO,Pernar LI,Apovian CM,Puri V,Gokce N
PA5-16754 was used in Western Blotting to support a key role and functional significance of FSP27 as a critical endogenous modulator of human microvascular function that has not been previously described.
Tue Jun 04 00:00:00 EDT 2019
Fat-Specific Protein 27 Regulation of Vascular Function in Human Obesity.
Journal of the American Heart Association
Karki S,Farb MG,Sharma VM,Jash S,Zizza EJ,Hess DT,Carmine B,Carter CO,Pernar LI,Apovian CM,Puri V,Gokce N
PA5-16754 was used in Western Blotting to support a key role and functional significance of FSP27 as a critical endogenous modulator of human microvascular function that has not been previously described.
Tue Jun 04 00:00:00 EDT 2019
Status of VEGF in preeclampsia and its effect on endoplasmic reticulum stress in placental trophoblast cells.
European journal of obstetrics & gynecology and reproductive biology: X
Mochan S,Dhingra MK,Gupta SK,Saxena S,Arora P,Yadav V,Rani N,Luthra K,Dwivedi S,Bhatla N,Dhingra R
Published figure using VEGF polyclonal antibody (Product # PA5-16754) in Immunohistochemistry
Tue Oct 01 00:00:00 EDT 2019
Modulation of cardiac vascular endothelial growth factor and PGC-1α with regular postexercise cold-water immersion of rats.
Journal of applied physiology (Bethesda, Md. : 1985)
Al-Horani RA,Al-Trad B,Haifawi S
PA5-16754 was used in Western Blotting to investigate the effects of postexercise CWI on the mitochondrial biogenesis (PGC-1), angiogenesis (VEGF), and cardioprotection (Hsp70 and SOD) factors in the myocardium.
Mon Apr 01 00:00:00 EDT 2019
Effects of excess thromboxane A2 on placental development and nutrient transporters in a Mus musculus model of fetal growth restriction.
Biology of reproduction
Gibbins KJ,Gibson-Corley KN,Brown AS,Wieben M,Law RC,Fung CM
PA5-16754 was used in Western Blotting to conclude that early-to-mid gestation hypertensive placentae show compensatory mechanisms to preserve fetal growth by increasing placental efficiencies and maintaining abundance of important nutrient transporters.
Tue May 01 00:00:00 EDT 2018
The role of miR-24 as a race related genetic factor in prostate cancer.
Oncotarget
Hashimoto Y,Shiina M,Kato T,Yamamura S,Tanaka Y,Majid S,Saini S,Shahryari V,Kulkarni P,Dasgupta P,Mitsui Y,Sumida M,Deng G,Tabatabai L,Kumar D,Dahiya R
PA516754 was used in western blot to suggest that miR-24 is a central regulator of race-related prostate cancer
Tue Mar 07 00:00:00 EST 2017
Endocardial-to-mesenchymal transformation and mesenchymal cell colonization at the onset of human cardiac valve development.
Development (Cambridge, England)
Monaghan MG,Linneweh M,Liebscher S,Van Handel B,Layland SL,Schenke-Layland K
PA5-16754 was used in immunohistochemistry - paraffin section to characterize human cardiac valve development by endocardial-to-mesenchymal transformation and mesenchymal cell colonization
Mon Feb 01 00:00:00 EST 2016
Deletion of IL-33R attenuates VEGF expression and enhances necrosis in mammary carcinoma.
Oncotarget
Milosavljevic MZ,Jovanovic IP,Pejnovic NN,Mitrovic SL,Arsenijevic NN,Simovic Markovic BJ,Lukic ML
PA5-16754 was used in immunohistochemistry - paraffin section to study the attenuation of VEGF expression and enhancement of necrosis in mammary carcinoma by deletion of IL-33R
Tue Apr 05 00:00:00 EDT 2016
Combined use of COX-1 and VEGF immunohistochemistry refines the histopathologic prognosis of renal cell carcinoma.
International journal of clinical and experimental pathology
Osman WM,Youssef NS
Published figure using VEGF polyclonal antibody (Product # PA5-16754) in Immunohistochemistry
Tue Jun 07 00:00:00 EDT 2016
Tumor endothelium FasL establishes a selective immune barrier promoting tolerance in tumors.
Nature medicine
Motz GT,Santoro SP,Wang LP,Garrabrant T,Lastra RR,Hagemann IS,Lal P,Feldman MD,Benencia F,Coukos G
PA5-16754 was used in immunohistochemistry to study the rmechanism by which tumor endothelial FasL promotes tumor immune tolerance
Sun Jun 01 00:00:00 EDT 2014
Profilin-1 phosphorylation directs angiocrine expression and glioblastoma progression through HIF-1α accumulation.
Nature cell biology
Fan Y,Potdar AA,Gong Y,Eswarappa SM,Donnola S,Lathia JD,Hambardzumyan D,Rich JN,Fox PL
PA5-16754 was used in western blot to study the roles of angiocrine secretion and accumulation of HIF-1alpha in the mechanism by which phosphorylation of endothelial cell profilin-1 promotes the progression of glioblastoma
Thu May 01 00:00:00 EDT 2014
Aging-related changes of optic nerve of Wistar albino rats.
Age (Dordrecht, Netherlands)
El-Sayyad HI,Khalifa SA,El-Sayyad FI,Al-Gebaly AS,El-Mansy AA,Mohammed EA
PA5-16754 was used in immunohistochemistry to study age-related deterioration of the optic nerve in a rat model
Tue Apr 01 00:00:00 EDT 2014
Castration resistant prostate cancer is associated with increased blood vessel stabilization and elevated levels of VEGF and Ang-2.
The Prostate
Tomić TT,Gustavsson H,Wang W,Jennbacken K,Welén K,Damber JE
PA5-16754 was used in immunohistochemistry to study blood vessel stabilization and the expression of pro-angiogenic VEGF and Ang-2 in castration-resistant prostate cancer
Tue May 15 00:00:00 EDT 2012
Proliferation and maturation of microvessels in arteriovenous malformations--expression patterns of angiogenic and cell cycle-dependent factors.
Journal of cutaneous pathology
Meijer-Jorna LB,van der Loos CM,Teeling P,de Boer OJ,Florquin S,van der Horst CM,van der Wal AC
PA5-16754 was used in immunohistochemistry to study the patterns of proliferation and maturation of microvessels in arteriovenous malformations
Fri Jun 01 00:00:00 EDT 2012
Inflammation modulates expression of laminin in the central nervous system following ischemic injury.
Journal of neuroinflammation
Ji K,Tsirka SE
PA5-16754 was used in western blot to study the role of COX-2 in the upregulation of CNS laminin in a murine model of ischemic injury
Tue Jul 03 00:00:00 EDT 2012