Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is expected to recognize both human isoforms of this protein.
It is predicted to react with bovine, canine and porcine based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to microtubular structures in the cytoplasm. Alternate splicing of this gene results in two transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ37715; FLJ41813; midin 2; Midin-2; Midline defect 2; Midline-2; Probable E3 ubiquitin-protein ligase MID2; RING finger protein 60; RING-type E3 ubiquitin transferase MID2; RP6-191P20.2; tripartite motif protein 1; Tripartite motif-containing protein 1
Gene Aliases: FXY2; MID2; MRX101; RNF60; TRIM1
UniProt ID: (Human) Q9UJV3
Entrez Gene ID: (Human) 11043
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support