Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Toll-like receptors (TLRs) are signaling molecules that recognize different microbial products during infection and serve as an important link between the innate and adaptive immune responses. These proteins act through adaptor molecules such as TIRAP and MyD88 to activate various kinases and transcription factors. In TIRAP-deficient mice, TLR signaling in response to TLR2 ligands (using either TLR1 and TLR6 as co-receptors) is totally abolished, suggesting that MyD88 and TIRAP work together and are both required for TLR2 signaling. Furthermore, these mice are also resistant to the toxic effects of LPS and show defects in NF-βB and MAP kinase activation, suggesting that TIRAP is also involed in TLR4 signaling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adapter protein wyatt; Adaptor protein Wyatt; FLJ42305; HGNC:17192; MyD88 adapter-like protein; MyD88-2; OTTHUMP00000179130; TIR domain-containing adapter protein; toll-interleukin 1 receptor (TIR) domain containing adaptor protein; Toll-like receptor adaptor protein; Toll/interleukin-1 receptor domain-containing adapter protein
Gene Aliases: BACTS1; MAL; MyD88-2; TIRAP; wyatt
UniProt ID: (Human) P58753
Entrez Gene ID: (Human) 114609
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support