Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Despite its predicted molecular weight, NADE often migrates at ~23kDa in SDS-PAGE. A suggested positive control is human brain tissue lysate.
PA5-20076 can be used with blocking peptide PEP-0194.
May be a signaling adapter molecule involved in p75NTR-mediated apoptosis induced by NGF. Plays a role in zinc-triggered neuronal death. May play an important role in the pathogenesis of neurogenetic diseases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brain expressed X-linked 3; Brain-expressed X-linked protein 3; Brain-expressed X-linked protein 3 homolog; granule cell antiserum positive 27; nerve growth factor receptor (TNFRSF16) associated protein 1; Nerve growth factor receptor-associated protein 1; Ovarian granulosa cell 13.0 kDa protein HGR74; p75NTR-associated Cell Death Executor; Protein BEX3; RP13-349O20.1
Gene Aliases: AL033356; Bex; BEX3; DXS6984E; DXWsu67e; Gcap27; HGR74; NADE; NGFRAP1
UniProt ID: (Human) Q00994, (Mouse) Q9WTZ9
Entrez Gene ID: (Human) 27018, (Mouse) 12070
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support