Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
A suggested positive control is K562 cell lysate.
PA5-20089 can be used with blocking peptide PEP-0207.
The PA5-20089 immunogen is located within amino acids 80-130 of BIM.
Predicted molecular ~ 22kD. In Western blot applications, this antibody has been observed to detect a band at: 22kD.
Predicted species reactivity based on immunogen sequence: Rat: (100%).
BIM belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. The protein encoded by this gene contains a Bcl-2 homology domain 3 (BH3). It has been shown to interact with other members of the BCL-2 protein family, including BCL2, BCL2L1/BCL-X(L), and MCL1, and to act as an apoptotic activator. The expression of this gene can be induced by nerve growth factor (NGF), as well as by the forkhead transcription factor FKHR-L1, which suggests a role of this gene in neuronal and lymphocyte apoptosis. Transgenic studies of the mouse counterpart suggested that this gene functions as an essential initiator of apoptosis in thymocyte-negative selection. Several alternatively spliced transcript variants of this gene have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bcl-2 interacting mediator of cell death; bcl-2 interacting protein Bim; Bcl-2 like 11; Bcl-2-like protein 11; bcl-2-related ovarian death agonist; Bcl2 interacting mediator of cell death; Bcl2-interacting mediator of cell death; Bcl2-L-11; BCL2-like 11 (apoptosis facilitator); Bim EL; Bim-A; BIM-alpha6; BIM-beta6; BIM-beta7; OTTHUMP00000197013; OTTHUMP00000204247
Gene Aliases: 1500006F24Rik; BAM; bcl2-L-11; BCL2L11; BIM; BOD
UniProt ID: (Human) O43521, (Mouse) O54918
Entrez Gene ID: (Human) 10018, (Mouse) 12125
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support