Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is Jurkat cell lysate.
PA5-20526 can be used with blocking peptide PEP-0646.
Epidermodysplasia verruciformis (EV) is an autosomal recessive genodermatosis associated with a high risk of skin cancers resulting from a high susceptibility to infection by specific human papillomaviruses. Mutations in two homologous genes EVER1 and EVER2 cause the majority of EV cases. These two proteins form a complex and interact with the zinc transporter ZnT-1 in the endoplasmic reticulum. Cells lacking EVER2 accumulated higher levels of zinc in the nucleolus and nucleus compare to those cells with and intact EVER2 gene, indicating that one role of EVER2 is to regulate the intracellular distribution of zinc.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epidermodysplasia verruciformis 2; Epidermodysplasia verruciformis protein 2; Epidermodysplasia verruciformis protein 2 homolog; transmembrane channel-like 8; Transmembrane channel-like protein 8
Gene Aliases: EV2; EVER2; EVIN2; mFLJ00400; TMC8
UniProt ID: (Human) Q8IU68, (Mouse) Q7TN58
Entrez Gene ID: (Human) 147138, (Mouse) 217356, (Rat) 688276
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support