Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Recommended positive controls: 293T, A431.
Predicted reactivity: Mouse (97%), Rat (97%), Pig (100%), Chicken (92%), Rhesus Monkey (100%), Bovine (98%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are produced by RNA polymerase II and are components of the heterogeneous nuclear RNA (hnRNA) complexes. They are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene, which binds to one of the components of the multiprotein editosome complex, has two repeats of quasi-RRM (RNA recognition motif) domains that bind to RNAs. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABBP-1; apobec-1 binding protein 1; APOBEC1-binding protein 1; apolipoprotein B mRNA editing enzyme, catalytic polypeptide 1-binding protein 1; Heterogeneous nuclear ribonucleoprotein A/B; hnRNP A/B; hnRNP type A/B protein
Gene Aliases: ABBP1; HNRNPAB; HNRPAB
UniProt ID: (Human) Q99729
Entrez Gene ID: (Human) 3182
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support