Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji.
Predicted reactivity: Mouse (80%), Pig (95%), Rhesus Monkey (100%), Bovine (92%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May be a signaling adapter molecule involved in p75NTR-mediated apoptosis induced by NGF. Plays a role in zinc-triggered neuronal death. May play an important role in the pathogenesis of neurogenetic diseases.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain-expressed X-linked protein 3; nerve growth factor receptor (TNFRSF16) associated protein 1; Nerve growth factor receptor-associated protein 1; Ovarian granulosa cell 13.0 kDa protein HGR74; p75NTR-associated cell death executor; Protein BEX3; RP13-349O20.1
Gene Aliases: Bex; BEX3; DXS6984E; HGR74; NADE; NGFRAP1
UniProt ID: (Human) Q00994
Entrez Gene ID: (Human) 27018
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support