Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 13
Recommended positive controls: A549, H1299, HCT116, HUVEC, Mouse heart, Rat bladder, CTGF-transfected 293T, HepG2, HepG2 conditioned medium.
Predicted reactivity: Mouse (93%), Rat (94%), Xenopus laevis (85%), Pig (95%), Chicken (94%), Sheep (96%), Chimpanzee (99%), Bovine (93%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Connective Tissue Growth Factor (CTGF), also called CCN2, is a member of the CCN family. CTGF is a secreted protein produced by umbilical veins and vascular endothelial cells. CTGF possesses an Insulin-like Growth Factor (IGF)-binding domain, a thrombospondin type 1 domain, and a cysteine knot region. CTGF plays important roles in the proliferation and differentiation of chondrocytes, induces angiogenesis, and promotes cell adhesion in fibroblasts, endothelial, and epithelial cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-Nov; CCN family member 2; Cellular communication network factor 2; Connective tissue growth factor; Connective tissue growth factor precursor (CTGF) (FISP-12 protein) (Hypertrophic chondrocyte-specific protein 24); Connective tissue growth-related protein; fibroblast inducible secreated protein; fibroblast inducible secreted protein; H-CTGF; hypertrophic chondrocyte-specific gene product 24; Hypertrophic chondrocyte-specific protein 24; IBP-8; IGF-binding protein 8; IGFBP-8; Insulin-like growth factor-binding protein 8; MGC102839; Protein FISP-12
Gene Aliases: betaIG-M2; CCN2; CTGF; CTGRP; Fisp-12; Fisp12; HCS24; IGFBP8; NOV2
UniProt ID: (Bovine) O18739, (Human) P29279, (Mouse) P29268, (Rat) Q9R1E9
Entrez Gene ID: (Bovine) 281103, (Human) 1490, (Mouse) 14219, (Rat) 64032
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support