Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
A suggested positive control for Western blot is JK cells.
These proteins belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. This gene encodes a protein that binds ephrin-A ligands.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain-specific kinase; CEK-7; eck-like sequence 1; EHK-1; EK7; EPH homology kinase 1; EPH-like kinase 7; Ephrin A2; Ephrin A5; Ephrin type-A receptor 2; Ephrin type-A receptor 5; EphrinA2; EphrinA5; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; receptor protein-tyrosine kinase HEK7; soluble EPHA2 variant 1; Tyrosine-protein kinase receptor ECK; tyrosine-protein kinase receptor EHK-1; Tyrosine-protein kinase receptor MPK-5; Tyrosine-protein kinase receptor SEK-2
Gene Aliases: AI854630; ARCC2; AW125296; AW545284; BSK; CEK7; CTPA; CTPP1; CTRCT6; ECK; EHK-1; EHK1; EK7; Els1; EPHA2; EPHA5; HEK7; Myk2; Rek7; Sek-2; Sek2; TYRO4
UniProt ID: (Human) B5A968, (Human) P54756, (Mouse) Q03145, (Mouse) Q60629
Entrez Gene ID: (Human) 1969, (Human) 2044, (Mouse) 13836, (Mouse) 13839
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support