Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: MSRSFYVDSL IIKDTSRPAP SLPEPHPGPD FFIPLGMPPP LVMSVSGPGC
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 93%
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Genetic-screened homeobox 2; GS homeobox 2; Homeobox protein GSH-2
Gene Aliases: GSH2; GSX2
UniProt ID: (Human) Q9BZM3
Entrez Gene ID: (Human) 170825
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support