Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: KMSGDVADST DARSTLSQVE PGNDRKGLDF NRQIKTEDLS DSLQQTLSHR
Sequence homology: Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 77%
This gene encodes a member of the POU domain family of transcription factors. POU domain transcription factors bind to a specific octamer DNA motif and regulate cell type-specific differentiation pathways. The encoded protein is primarily expressed in the epidermis, and plays a critical role in keratinocyte proliferation and differentiation. The encoded protein is also a candidate tumor suppressor protein, and aberrant promoter methylation of this gene may play a role in cervical cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. [provided by RefSeq, Sep 2011].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 11-Oct; Epoc-1; FLJ40063; MGC126698; Oct-11; Octamer-binding protein 11; Octamer-binding transcription factor 11; OTF-11; POU domain transcription factor OCT11a; POU domain, class 2, transcription factor 3; Rov 1 pou-homeodomain protein; Transcription factor PLA-1; Transcription factor Skn-1
Gene Aliases: Epoc-1; Epoc1; OCT-11; Oct-11a; OCT11; OTF-11; OTF11; PLA-1; PLA1; POU2F3; Skin; Skin-1a; Skn-1; Skn-1a; Skn-li; Skn1
UniProt ID: (Human) Q9UKI9, (Rat) P42571, (Mouse) P31362
Entrez Gene ID: (Human) 25833, (Rat) 116544, (Mouse) 18988
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support