Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
Allergin 1 is a protein coding gene. Immunoglobulin-like receptor which plays an inhibitory role in degranulation of mast cells. Negatively regulates IgE-mediated mast cell activation and suppresses the type I immediate hypersensitivity reaction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Allergin-1; Allergy inhibitory receptor 1; Mast cell antigen 32; Mast cell immunoglobulin-like receptor 1; MCA-32; MCA32-1S2; probable mast cell antigen 32 homolog
Gene Aliases: Allergin-1; C17orf60; MCA-32; MCA32; MILR1
UniProt ID: (Human) Q7Z6M3
Entrez Gene ID: (Human) 284021
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support