Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody detects endogenous levels of total ALG12 protein.
This gene encodes a member of the glycosyltransferase 22 family. The encoded protein catalyzes the addition of the eighth mannose residue in an alpha-1,6 linkage onto the dolichol-PP-oligosaccharide precursor (dolichol-PP-Man(7)GlcNAc(2)) required for protein glycosylation. Mutations in this gene have been associated with congenital disorder of glycosylation type Ig (CDG-Ig)characterized by abnormal N-glycosylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: asparagine-linked glycosylation 12 homolog (S. cerevisiae, alpha-1,6-mannosyltransferase); asparagine-linked glycosylation 12 homolog (yeast, alpha-1,6-mannosyltransferase); asparagine-linked glycosylation 12, alpha-1,6-mannosyltransferase homolog; Asparagine-linked glycosylation protein 12 homolog; dol-P-Man dependent alpha-1,6-mannosyltransferase; Dol-P-Man:Man(7)GlcNAc(2)-PP-Dol alpha-1,6-mannosyltransferase; dolichyl-P-Man:Man(7)GlcNAc(2)-PP-dolichol alpha-1,6-mannosyltransferase; Dolichyl-P-Man:Man(7)GlcNAc(2)-PP-dolichyl-alpha-1,6-mannosyltransferase; dolichyl-P-mannose:Man-7-GlcNAc-2-PP-dolichyl-alpha-6-mannosyltransferase; hALG12; Mannosyltransferase ALG12 homolog; Membrane protein SB87
Gene Aliases: ALG12; CDG1G; ECM39; hALG12; PP14673
UniProt ID: (Human) Q9BV10
Entrez Gene ID: (Human) 79087
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support