Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LAVLLVLFLH IKNLPAQAAD TCPEVKVVGL EGSDKLTILR GCPGLPGAPG PKGEAGVIGE RGERGLPGAP GKAGPVGPKG DRGEKGMRGE KGDAGQSQSC ATGPRNCKD
Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 75%.
M-Ficolin (Ficolin-1) is a non-serum protein that is expressed in leukocytes and lung. M-Ficolin precipitates with mannose-binding lectin (MBL)-associated serine proteases (MASP)-1 and MASP-2, indicating that M-Ficolin forms complexes with MASP-1 and MASP-2, recognizes certain types of bacteria and subsequently activates the lectin pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Collagen/fibrinogen domain-containing protein 1; Ficolin-1; Ficolin-A; Ficolin-alpha; M-ficolin
Gene Aliases: FCN1; FCNM
UniProt ID: (Human) O00602
Entrez Gene ID: (Human) 2219
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support