Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Immunogen sequence: DVLSSNEATM QSMESLLRAK GLEVEQLSTT CQNLQWLKEE METKFSRWQK EQESIIQQLQ TSLHDRNKEV EDLSATLLCK LGPGQSEIAE ELCQRLQRKE RMLQDLLSDR NKQVLEHEME IQGLLQSVST REQESQAAA
Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 87%.
The protein encoded by this gene serves to anchor phosphodiesterase 4D to the Golgi/centrosome region of the cell. Defects in this gene may be a cause of myeloproliferative disorder (MBD) associated with eosinophilia. Several transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cardiomyopathy-associated protein 2; Myomegalin; myomegalin/phosphodiesterase 4D interacting protein variant 8; Phosphodiesterase 4D-interacting protein; Phosphodiesterase-binding protein clone 46; upregulated during skeletal muscle growth 4
Gene Aliases: 4732458A06Rik; 9430063L05Rik; C87016; CMYA2; D130016K21Rik; D3Bwg1078e; KIAA0454; KIAA0477; mKIAA0454; MMGL; Pbp46; PDE4DIP; Usmg4
UniProt ID: (Human) Q5VU46, (Rat) Q9WUJ3, (Mouse) Q80YT7
Entrez Gene ID: (Human) 9659, (Rat) 64183, (Mouse) 83679
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support