Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 1
Immunogen sequence: KLNLQVLLSH VLGKDVPRDG KAEFACSKCA FMLDRIYRFD TVIARIEALS IERLQKLLLE KDRLKFCIAS MYRKNNDDSG AEIKAGNGTV DMSVLPDARY SALLQEDFAY SGFECWVENE DQIQEPHSCH GSEGPGNRPR RCRGCA
Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 19%.
The protein encoded by this gene serves to anchor phosphodiesterase 4D to the Golgi/centrosome region of the cell. Defects in this gene may be a cause of myeloproliferative disorder (MBD) associated with eosinophilia. Several transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support