Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 8
Immunogen sequence: LTGLSLLPLS EKAARARQEE LYSELQARET FEKTPVEVPV GGFKGRTVTV WELISSEYFT AEQRQELLRQ FRTGKVTVEK VIKILIT
Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 91%.
Plectin is a prominent member of an important family of structurally and in part functionally related proteins, termed plakins or cytolinkers, that are capable of interlinking different elements of the cytoskeleton. Plakins, with their multi-domain structure and enormous size, not only play crucial roles in maintaining cell and tissue integrity and orchestrating dynamic changes in cytoarchitecture and cell shape, but also serve as scaffolding platforms for the assembly, positioning, and regulation of signaling complexes. Plectin is expressed as several protein isoforms in a wide range of cell types and tissues from a single gene located on chromosome 8 in humans. The expression of specific plectin isoforms was found to be dependent on cell type (tissue) and stage of development and it appears that each cell type (tissue) contains a unique set (proportion and composition) of plectin isoforms which carry out distinct and specific functions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HD1; Hemidesmosomal protein 1; PCN; Plectin; plectin 1, intermediate filament binding protein 500kDa; Plectin-1
Gene Aliases: EBS1; EBSMD; EBSND; EBSO; EBSOG; EBSPA; HD1; LGMD2Q; PCN; PLEC; PLEC1; PLEC1b; PLTN
UniProt ID: (Human) Q15149
Entrez Gene ID: (Human) 5339
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support