Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DSLGREAKCY NELNGCTKIY DPVCGTDGNT YPNECVLCFE NRKRQTSILI Q
Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 65%.
This is a trypsin inhibitor, its physiological function is to prevent the trypsin-catalyzed premature activation of zymogens within the pancreas. Defects in SPINK1 are a cause of pancreatitis (PCTT) [MIM:167800]. A disease characterized by the presence of calculi in pancreatic ducts. It causes severe abdominal pain attacks. Defects in SPINK1 are the cause of susceptibility to tropical calcific pancreatitis (TCP) [MIM:608189]. TCP is an idiopathic, juvenile, nonalcoholic form of chronic pancreatitis widely prevalent in several tropical countries. It can be associated with fibrocalculous pancreatic diabetes (FCPD) depending on both environmental and genetic factors. TCP differs from alcoholic pancreatitis by a much younger age of onset, pancreatic calcification, a high incidence of insulin dependent but ketosis resistant diabetes mellitus, and an exceptionally high incidence of pancreatic cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ISK1; Pancreatic secretory trypsin inhibitor; Serine protease inhibitor Kazal-type 1; serine protease inhibitor, Kazal type 1; Spink 3; TATI; Tumor-associated trypsin inhibitor
Gene Aliases: PCTT; PSTI; SPINK1; Spink3; TATI; TCP
UniProt ID: (Human) P00995
Entrez Gene ID: (Human) 6690
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support