Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KFTCHYRCRA LVCLDCCGPR DLGWEPAVER DTNVDEPVEW ETPDLSQAEI EQKIKEYNAQ INSNLFMSLN KDG
Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.
This gene encodes a protein similar to the RAS effector proteins. Loss or altered expression of this gene has been associated with the pathogenesis of a variety of cancers, which suggests the tumor suppressor function of this gene. The inactivation of this gene was found to be correlated with the hypermethylation of its CpG-island promoter region. The encoded protein was found to interact with DNA repair protein XPA. The protein was also shown to inhibit the accumulation of cyclin D1, and thus induce cell cycle arrest. Seven alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cardiac-specific ras association domain family 1 protein; pancreas-specific ras association domain family 1 protein; Ras association (RalGDS/AF-6) domain family member 1; Ras association domain-containing protein 1; RASSF1; tumor suppressor protein RDA32; WUGSC:H_LUCA12.5
Gene Aliases: 123F2; NORE2A; RASSF1; RASSF1A; RDA32; REH3P21
UniProt ID: (Human) Q9NS23
Entrez Gene ID: (Human) 11186
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support