Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PSFWILARAL KEFVAKEGQG NLPVRGTIPD MIADSGKYIK LQNVYREKAK KDAAAVGNHV AKLLQSIGQA PESISEKELK LLCSNSAFLR VVRCRSLAEE YGLDTINKDE
Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
The protein encoded by this gene binds to the beta-amyloid precursor protein. Beta-amyloid precursor protein is a cell surface protein with signal-transducing properties, and it is thought to play a role in the pathogenesis of Alzheimer's disease. In addition, the encoded protein can form a heterodimer with UBE1C and bind and activate NEDD8, a ubiquitin-like protein. This protein is required for cell cycle progression through the S/M checkpoint. Three transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: amyloid beta precursor protein binding protein 1, 59kDa; Amyloid beta precursor protein-binding protein 1, 59 kDa; amyloid beta precursor protein-binding protein 1, 59kD; Amyloid protein-binding protein 1; APP-BP1; NEDD8-activating enzyme E1 regulatory subunit; NEDD8-activating enzyme E1 subunit; Proto-oncogene protein 1; protooncogene protein 1
Gene Aliases: 59kDa; A-116A10.1; APPBP1; HPP1; NAE1; ula-1
UniProt ID: (Human) Q13564, (Mouse) Q8VBW6
Entrez Gene ID: (Human) 8883, (Mouse) 234664, (Rat) 84019
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support