Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NPRAAELFHQ LSQALEVLTD AAARAAYDKV RKAKKQAAER TQKLDEKRKK VKLDLEARER QAQAQES
Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
May negatively affect PAX8-induced thyroglobulin/TG transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DnaJ (Hsp40) homolog, subfamily C, member 17; DnaJ homolog subfamily C member 17
Gene Aliases: DNAJC17
UniProt ID: (Human) Q9NVM6
Entrez Gene ID: (Human) 55192
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support