Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKTLKSQMGT LVEKGRHDDE LIDALMDQLK QLQEILGSLS LQEEKTRVSQ HHLDQQLNSE AQRSNSLVAQ LQAMVAEREA KVRQLEMEIG QLNVHYLRN
Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 81%.
Required for primary cilia formation and promotes the localization of the ciliopathy protein BBS4 to both centriolar satellites and cilia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support