Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EHSNGTVAPD SPASPASDGP ALPSPAIPRG GFALSLLANA E
Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.
This gene encodes a protein with a leucine-rich repeat and a calponin homology domain. Polymorphism in this gene may be associated with susceptibililty to knee osteoarthritis. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: JP-2; Junctophilin type 2; Junctophilin-2
Gene Aliases: CMH17; JP-2; JP2; JPH2
UniProt ID: (Human) Q9BR39
Entrez Gene ID: (Human) 57158
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support