Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Immunogen sequence: KVREESRAQS PVEAEFPYGQ LSTSCHSHLV PQNGHQATHL EVTKQPTNRV
Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 56%.
This gene encodes a membrane-bound glycoprotein from the family of dipeptidases involved in hydrolytic metabolism of various dipeptides, including penem and carbapenem beta-lactam antibiotics. This gene is located on chromosome 16 in a cluster with another member of this family. Alternatively spliced transcript variants that encode different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dipeptidase 3; membrane-bound dipeptidase 3; metallopeptidase (family M19); testis secretory sperm-binding protein Li 211a
Gene Aliases: DPEP3; MBD3; UNQ834/PRO1772
UniProt ID: (Human) Q9H4B8
Entrez Gene ID: (Human) 64180
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support