Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 79%; Dog: 92%; Horse: 92%; Human: 100%; Rabbit: 83%
Protein which interacts with the E3 UFM1-protein ligase UFL1 and one of its substrates TRIP4 and is required for TRIP4 ufmylation. Through TRIP4 ufmylation may regulate nuclear receptors-mediated transcription. May play a role in NF-kappa-B-mediated transcription through regulation of the phosphorylation and the degradation of NFKBIA, the inhibitor of NF-kappa-B. May also play a role in the cellular response to endoplasmic reticulum stress.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dashurin; DDRGK domain-containing protein 1; UFM1-binding and PCI domain-containing protein 1; UFM1-binding protein 1 containing a PCI domain
Gene Aliases: C20orf116; DDRGK1; dJ1187M17.3; UFBP1
UniProt ID: (Human) Q96HY6
Entrez Gene ID: (Human) 65992
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support