Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive Control: mouse heart
Predicted Reactivity: Human (100%), Rat (91%), Rhesus Monkey (99%), Bovine (91%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PRELP is a leucine-rich repeat protein present in connective tissue extracellular matrix. This protein functions as a molecule anchoring basement membranes to the underlying connective tissue. This protein has been shown to bind type I collagen to basement membranes and type II collagen to cartilage. It also binds the basement membrane heparan sulfate proteoglycan perlecan. This protein is suggested to be involved in the pathogenesis of Hutchinson-Gilford progeria (HGP), which is reported to lack the binding of collagen in basement membranes and cartilage.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 55 kDa leucine-rich repeat protein of articular cartilage; Prolargin; prolargin proteoglycan; Proline-arginine-rich end leucine-rich repeat protein; proline/arginine-rich end leucine-rich repeat protein
Gene Aliases: 7330409J17Rik; MST161; MSTP161; PRELP; SLRR2A
UniProt ID: (Human) P51888, (Mouse) Q9JK53
Entrez Gene ID: (Human) 5549, (Mouse) 116847
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support