Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
TARC ligand (CCL17) belongs to the CC chemokine family. It is a small cytokine and the gene is located on q arm of chromosome 16, in humans, along with other chemokines. TARC specifically binds to T-cells and interacts with chemokine receptors CCR4 and CCR8. It plays an important role in T cell development in thymus as well as in trafficking and activation of mature T cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABCD-2; C-C motif chemokine; C-C motif chemokine 17; CC chemokine TARC; small inducible cytokine subfamily A17; Small-inducible cytokine A17; Thymus and activation-regulated chemokine
Gene Aliases: Abcd-2; Ccl17; Scya17; Scya17l; Tarc
UniProt ID: (Mouse) Q9WUZ6
Entrez Gene ID: (Mouse) 20295
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support