Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
EPHB2 is a receptor tyrosine kinase that belongs to the ephrin receptor family. Members of the Eph family of kinases play important roles in diverse biological processes including nervous system development, angiogenesis and neural synapsis formation and maturation. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. EphB4 binds to ephrin-B2 and plays an essential role in vascular development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Developmentally-regulated Eph-related tyrosine kinase; EFL6; EFNB3; EK5; ELK-related tyrosine kinase; EPH B2; EPH tyrosine kinase 3; EPH-like kinase 5; EPHB3; Ephrin B1; Ephrin B2; Ephrin B3; Ephrin B4; Ephrin type-B receptor 2; EphrinB1; EphrinB2; EphrinB3; EphrinB4; EPLG8; ETK2; HEK-6; HEK2; LERK8; MGC87492; Neural kinase; Nuk receptor tyrosine kinase; protein-tyrosine kinase HEK5; Renal carcinoma antigen NY-REN-47; TYRO6; Tyrosine-protein kinase receptor EPH-3; Tyrosine-protein kinase receptor SEK-3; Tyrosine-protein kinase TYRO5
Gene Aliases: CAPB; Cek5; DRT; EK5; EPHB2; EPHT3; EPTH3; ERK; ETECK; HEK5; Nuk; PCBC; Prkm5; Qek5; Sek3; TYRO5
UniProt ID: (Human) P29323, (Mouse) P54763
Entrez Gene ID: (Human) 2048, (Mouse) 13844
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support