Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Immunogen sequence: REEEKPFRDC ADVYQAGFNK SGIYTIYINN MPEPKKVFCN MDVNGGGWTV IQHREDGSLD FQRGWKEYKM GFGNPSGEYW LGNEFIFAIT SQRQYMLRIE LMDWEGNRAY SQYDRFHIGN EKQNYRLYLK GHTGTAGKQS SLILHGADFS TKDADNDNCM CKCALMLTGG WWFDACGPSN LNGMFYTAGQ NHGKLNGIKW HYFKGPSYSL RSTTMMIRPL DF
Positive Samples: A549, mouse lung
Angiopoietins are proteins with important roles in vascular development and angiogenesis. All angiopoietins bind with similar affinity to an endothelial cell-specific tyrosine-protein kinase receptor. The protein encoded by this gene is a secreted glycoprotein that activates the receptor by inducing its tyrosine phosphorylation. It plays a critical role in mediating reciprocal interactions between the endothelium and surrounding matrix and mesenchyme. The protein also contributes to blood vessel maturation and stability, and may be involved in early development of the heart.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ANG-1; Angiopoietin-1
Gene Aliases: 1110046O21Rik; AGP1; AGPT; Agpt1; Ang-1; ANG1; ANGPT1; KIAA0003
UniProt ID: (Human) Q15389, (Mouse) O08538, (Rat) O35460
Entrez Gene ID: (Human) 284, (Mouse) 11600, (Rat) 89807
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support